Skip to Content
Merck
All Photos(2)

Key Documents

A3882

Sigma-Aldrich

Alizarin-3-methyliminodiacetic acid

≥98% (TLC), powder

Synonym(s):

N-(carboxymethyl)-N-[(9,10-dihydro-3,4-dihydroxy-9,10-dioxo-2-anthracenyl)methyl]-glycine, 3,4-Dihydroxyanthraquinon-2-yl-methylimino-diacetic acid, Alizarin Complexone, Alizarin Fluorine Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15NO8
CAS Number:
Molecular Weight:
385.32
Beilstein:
2190028
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

Alizarin-3-methyliminodiacetic acid,

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

1 M NaOH: 10 mg/mL, blue to purple

SMILES string

OC(=O)CN(CC(O)=O)Cc1cc2C(=O)c3ccccc3C(=O)c2c(O)c1O

InChI

1S/C19H15NO8/c21-13(22)7-20(8-14(23)24)6-9-5-12-15(19(28)16(9)25)18(27)11-4-2-1-3-10(11)17(12)26/h1-5,25,28H,6-8H2,(H,21,22)(H,23,24)

InChI key

PWIGYBONXWGOQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Alizarin-3-methyliminodiacetic acid is used in the colorimetric detection of fluorine. It is a dye employed to stain mineralized bones of the preserved specimen from vertebrates.

Application

Alizarin-3-methyliminodiacetic acid has been used:
  • for live staining of mineralized bone matrix
  • to double-label the bone for dynamic bone histomorphometry at the femoral mid-diaphysis in the rat
  • as a label in histomorphometry of the tibial diaphysis to indicate new bone formation in the rat

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An evaluation of staining techniques for marking daily growth in scleractinian corals
Holcomb M, et al.
Journal of Experimental Marine Biology and Ecology, 440, 126-131 (2013)
A Bensimon-Brito et al.
BMC developmental biology, 16, 2-2 (2016-01-21)
The correct evaluation of mineralization is fundamental for the study of skeletal development, maintenance, and regeneration. Current methods to visualize mineralized tissue in zebrafish rely on: 1) fixed specimens; 2) radiographic and μCT techniques, that are ultimately limited in resolution;
J M Shand et al.
International journal of oral and maxillofacial surgery, 31(5), 525-531 (2002-11-07)
The purpose of this study was to investigate the incorporation of fresh frozen irradiated membranous allogeneic bone grafts into critical size calvarial defects in the rabbit. Fifteen rabbits had calvarial defects prepared. Twelve rabbits received allogeneic grafts and three received
Bi Foua Claude Alain Gohi et al.
Cell & bioscience, 10, 2-2 (2020-01-11)
A two-stage method of obtaining viable human amniotic stem cells (hAMSCs) in large-scale is described. First, human amniotic stem cells are isolated via dual enzyme (collagenase II and DNAase I) digestion. Next, relying on a culture of the cells from
Ling Zhu et al.
Dalton transactions (Cambridge, England : 2003), 48(15), 5035-5047 (2019-03-28)
A complexone analog chemosensor, H2L, bearing chelating bis(2-pyridylmethyl) amine and alizarin groups was synthesized via the Mannich reaction. H2L chromically responds to OH-, F-, CH3COO-, and H2PO4- in DMF, CH3CN, and acetone, but not in CH3OH or H2O. The addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service