Skip to Content
Merck
All Photos(1)

Key Documents

PZ0193

Sigma-Aldrich

Axitinib

≥98% (HPLC), powder, tyrosine kinase inhibitor

Synonym(s):

AG-013736, N-Methyl-2-((3-((1E)-2-(pyridin-2-yl)ethenyl)-1H-indazol-6-yl)sulfanyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N4OS
CAS Number:
Molecular Weight:
386.47
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Product Name

Axitinib, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥8 mg/mL

storage temp.

room temp

SMILES string

CNC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)n[nH]c3c2

InChI

1S/C22H18N4OS/c1-23-22(27)18-7-2-3-8-21(18)28-16-10-11-17-19(25-26-20(17)14-16)12-9-15-6-4-5-13-24-15/h2-14H,1H3,(H,23,27)(H,25,26)/b12-9+

InChI key

RITAVMQDGBJQJZ-FMIVXFBMSA-N

Gene Information

Related Categories

Application

Axitinib has been used as a vascular endothelial growth factor (VEGF) receptor tyrosine kinase inhibitor (TKI) to study its effect on renal cancer cell proliferation in vitro. It has also been used to inhibit anterior mesenteric artery (AMA) angiogenesis in dechlorinated embryos maintained in conditioned water (CW).

Biochem/physiol Actions

Axitinib (AG-013736) is an orally available, potent (picomolar) and selective tyrosine kinase inhibitor that blocks VEGF receptors 1, 2 and 3. The drug blocks VEGF-mediated endothelial cell survival, tube formation, and downstream signaling through endothelial nitric oxide synthase, Akt and extracellular signal-regulated kinase.
Orally available, potent and selective inhibitor of VEGF receptors 1, 2 and 3

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxic 3D in vitro culture models reveal distinct resistance processes to TKIs in renal cancer cells
Bielecka Z, et al.
Cell & Bioscience, 7(1), 71-71 (2017)
Ana Carolina Monteiro et al.
Molecular oncology, 13(6), 1433-1449 (2019-05-10)
The high mortality rate of melanoma is broadly associated with its metastatic potential. Tumor cell dissemination is strictly dependent on vascularization; therefore, angiogenesis and lymphangiogenesis play an essential role in metastasis. Hence, a better understanding of the players of tumor
Fatma Demircan Yildirim et al.
Medycyna pracy, 71(6), 649-663 (2020-10-28)
About 8 million healthcare workers in the USA are potentially exposed to hazardous drugs or their toxic metabolites over a long period of time despite the fact that both the Occupational Safety and Health Administration and the European Parliament recommend
Renin expression in developing zebrafish is associated with angiogenesis and requires the Notch pathway and endothelium
Rider S, et al.
American Journal of Physiology: Renal Physiology, 309(6), F531-F539 (2015)
Heidi I Chen et al.
The Journal of clinical investigation, 124(11), 4899-4914 (2014-10-02)
Coronary arteries (CAs) stem from the aorta at 2 highly stereotyped locations, deviations from which can cause myocardial ischemia and death. CA stems form during embryogenesis when peritruncal blood vessels encircle the cardiac outflow tract and invade the aorta, but

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service