Skip to Content
Merck
All Photos(1)

Documents

CDS023389

Sigma-Aldrich

Dasatinib

Synonym(s):

N-(2-Chloro-6-methylphenyl)-2-[[6-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-5-thiazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26ClN7O2S
CAS Number:
Molecular Weight:
488.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

storage temp.

-10 to -25°C

SMILES string

CC1=NC(NC2=NC=C(C(NC3=C(C=CC=C3Cl)C)=O)S2)=CC(N4CCN(CC4)CCO)=N1

InChI

1S/C22H26ClN7O2S/c1-14-4-3-5-16(23)20(14)28-21(32)17-13-24-22(33-17)27-18-12-19(26-15(2)25-18)30-8-6-29(7-9-30)10-11-31/h3-5,12-13,31H,6-11H2,1-2H3,(H,28,32)(H,24,25,26,27)

InChI key

ZBNZXTGUTAYRHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Gastro-intestinal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maja Kim Kuepper et al.
Leukemia, 33(8), 1964-1977 (2019-03-08)
Tyrosine kinase inhibitor (TKI) therapy effectively blocks oncogenic Bcr-Abl signaling and induces molecular remission in the majority of CML patients. However, the disease-driving stem cell population is not fully targeted by TKI therapy in the majority of patients, and leukemic
Kenta Mukaihara et al.
PloS one, 12(9), e0185321-e0185321 (2017-09-26)
Alveolar soft part sarcoma (ASPS) is an extremely rare metastatic soft tissue tumor with a poor prognosis for which no effective systemic therapies have yet been established. Therefore, the development of novel effective treatment approaches is required. Tyrosine kinases (TKs)
Lei Liu et al.
Journal of pineal research, 67(2), e12588-e12588 (2019-05-30)
Chordoma is an extremely rare malignant bone tumor with a high rate of relapse. While cancer stem cells (CSCs) are closely associated with tumor recurrence, which depend on its capacity to self-renew and induce chemo-/radioresistance, whether and how CSCs participate
Katherine Tarlock et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(16), 5038-5048 (2019-06-12)
KIT mutations (KIT+) are common in core binding factor (CBF) AML and have been associated with varying prognostic significance. We sought to define the functional and clinical significance of distinct KIT mutations in CBF pediatric AML. Following transfection of exon
Diahann T S L Jansen et al.
Frontiers in immunology, 9, 8-8 (2018-02-07)
With the advent of novel strategies to induce tolerance in autoimmune and autoimmune-like conditions, clinical trials of antigen-specific tolerizing immunotherapy have become a reality. Besides safety, it will be essential to gather mechanistic data on responding CD4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service