Skip to Content
Merck
All Photos(1)

Key Documents

P7965

Sigma-Aldrich

Monoclonal Anti-P-Glycoprotein (MDR) antibody produced in mouse

clone F4, ascites fluid

Synonym(s):

Anti-ABC20, Anti-CD243, Anti-CLCS, Anti-GP170, Anti-MDR1, Anti-P-GP, Anti-PGY1, Anti-p-170

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

F4, monoclonal

mol wt

antigen 170-180 kDa

contains

15 mM sodium azide

species reactivity

hamster, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500 using human kidney sections
immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
radioimmunoassay: suitable using cell-surface RIA
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCB1(5243)

General description

The ABCB1 (ATP binding cassette subfamily B member 1) gene is mapped to human chromosome 7q21.12. It encodes for P-glycoprotein, which is a membrane bound drug transporter protein, belonging to the ATP-binding cassette (ABC) transporters family. The gene is considered to be highly polymorphic.

Specificity

The antibody recognizes an epitope located in the amino terminal half of P-glycoprotein (Pgp), at the third extracellular loop of the molecule. The epitope is resistant to formalin fixation and periodate oxidation. The antibody detects specifically human MDR1 P-glycoprotein, but does not appear to recognize the human MDR3 product, nor the mouse mdr1a, mdr1b or the mdr3 P-glycoprotein.

Immunogen

mixture of human and hamster drug-resistant whole cells and crude plasma membranes.

Application

Monoclonal Anti-P-Glycoprotein (MDR) antibody produced in mouse has been used in western blot analysis and Immunohistochemistry.

Biochem/physiol Actions

Overexpression of ABCB1 (ATP binding cassette subfamily B member 1) gene is a major cause for multi-drug resistance, which makes chemotherapy challenging for the treatment of osteosarcoma. P-glycoprotein (P-gp) mediates the energy-dependent efflux of xenobiotic to the outside of plasma membrane from the inside of the cell , thereby affecting the process of absorption, distribution, and excretion of drugs. Cyclosporine, a calcineurin inhibitor serves as a substrate for P-gp.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T M Chu et al.
Biochemical and biophysical research communications, 203(1), 506-512 (1994-08-30)
Multidrug resistance (MDR) is a unique phenomenon in cancer patients and is commonly associated with an overexpression of the human MDR gene mdr1, which encodes an energy-dependent Mr 180 kDa membrane bound protein, known as P-glycoprotein. P-glycoprotein serves as a
T M Chu et al.
Hybridoma, 12(4), 417-429 (1993-08-01)
Using viable adriamycin resistant human ovarian carcinoma cells 2780AD and colchicine resistant human oral epidermoid carcinoma cells KB-24 as the immunogen in primary and subsequent i.p. immunizations, followed by i.v. boostings with crude plasma membranes of 2780AD, KB-24, Chinese hamster
Jingwei Ma et al.
Cell research, 26(6), 713-727 (2016-05-12)
Developing novel approaches to reverse the drug resistance of tumor-repopulating cells (TRCs) or stem cell-like cancer cells is an urgent clinical need to improve outcomes of cancer patients. Here we show an innovative approach that reverses drug resistance of TRCs
Di-2-pyridylketone 4, 4-Dimethyl-3-thiosemicarbazone (Dp44mT) Overcomes Multidrug-Resistance by a Novel Mechanism Involving the Hijacking of Lysosomal P-Glycoprotein (Pgp).
Jansson P J, et al.
The Journal of Biological Chemistry (2015)
Yun-Kai Zhang et al.
Scientific reports, 6, 25694-25694 (2016-05-10)
ATP-Binding Cassette transporters are involved in the efflux of xenobiotic compounds and are responsible for decreasing drug accumulation in multidrug resistant (MDR) cells. Discovered by structure-based virtual screening algorithms, bafetinib, a Bcr-Abl/Lyn tyrosine kinase inhibitor, was found to have inhibitory

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service