Skip to Content
Merck
All Photos(1)

Key Documents

P8920

Sigma-Aldrich

Poly-L-lysine solution

Poly-L-lysine solution
1 of 2 reviewers received a sample product or took part in a promotion

0.1 % (w/v) in H2O

Synonym(s):

2-(4-Cyanoanilino)-4,6-dichloropyrimidine, 4-[(4,6-Dichloro-2-pyrimidinyl)amino]benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116121
eCl@ss:
32160406
NACRES:
NA.47
Pricing and availability is not currently available.

biological source

synthetic

shelf life

Expiry date on the label.

mol wt

150,000-300,000

concentration

0.1 % (w/v) in H2O

color

colorless

application(s)

hematology
histology

storage temp.

room temp

SMILES string

N([C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)O)C(=O)[C@@H](N)CCCCN

InChI

1S/C18H38N6O4/c19-10-4-1-7-13(22)16(25)23-14(8-2-5-11-20)17(26)24-15(18(27)28)9-3-6-12-21/h13-15H,1-12,19-22H2,(H,23,25)(H,24,26)(H,27,28)/t13-,14-,15-/m0/s1

InChI key

WBSCNDJQPKSPII-KKUMJFAQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.
Poly-L-lysine solution has been used for the attachment of endothelial cell cross sections to UV-crosslinked PET membrane frame slides.[1] Poly-L-lysine coated slides were used for the placement of tissue sections for immunohistochemical assay.[2]

Biochem/physiol Actions

Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When it is absorbed to the cell culture surface, poly-L-lysine functions to increase the number of positively charged sites available for cell binding. With cells that can digest poly-L-lysine, poly-D-lysine should be used as the attachment factor.

Components

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Preparation Note

This product is supplied as a 0.10% (w/v) solution in water. Coating slides with this solution should be followed with a 5 minute incubation and drying the slides at room temperature. Coated slides will be stable for one year if protected from dust.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of the receptor activator for nuclear factor-?B ligand and osteoprotegerin in chronic otitis media.
Kuczkowski J et al.
American Journal of Otolaryngology, 31, 404-404 (2010)
Kimberly Cramer et al.
International journal of molecular sciences, 20(14) (2019-07-13)
In situ visualization of molecular assemblies near their macromolecular scale is a powerful tool to investigate fundamental cellular processes. Super-resolution light microscopies (SRM) overcome the diffraction limit and allow researchers to investigate molecular arrangements at the nanoscale. However, in bacterial
Hermann C Altmeppen et al.
Molecular neurodegeneration, 6, 36-36 (2011-05-31)
The cellular prion protein (PrPC) fulfils several yet not completely understood physiological functions. Apart from these functions, it has the ability to misfold into a pathogenic scrapie form (PrPSc) leading to fatal transmissible spongiform encephalopathies. Proteolytic processing of PrPC generates
Guojun Shi et al.
The Journal of clinical investigation, 127(10), 3897-3912 (2017-09-19)
Peptide hormones are crucial regulators of many aspects of human physiology. Mutations that alter these signaling peptides are associated with physiological imbalances that underlie diseases. However, the conformational maturation of peptide hormone precursors (prohormones) in the ER remains largely unexplored.
Daniel A Balikov et al.
Nucleus (Austin, Tex.), 8(5), 534-547 (2017-06-24)
The cytoskeleton provides structure and plays an important role in cellular function such as migration, resisting compression forces, and transport. The cytoskeleton also reacts to physical cues such as fluid shear stress or extracellular matrix remodeling by reorganizing filament associations

Articles

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

Human Protein Atlas uses A-431, U-251 MG, and U-205 cell lines with Prestige Antibodies for organelle mapping.

See All

Related Content

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Learn about the clinical study of blood, blood-forming organs, and blood diseases including the treatment, prevention, and stains and dyes used in hematology testing.

Questions

1–6 of 6 Questions  
  1. Can I add this to maxisorp plates surfaces to make DNA binding stronger.

    1 answer
    1. The Poly-L-Lysine solution (P8920) was developed and tested to improve tissue adhesion to glass slides for histological and immunohistochemical use. Although other uses of P8920 have been reported, there is no data on hand regarding the use of P8920 to make the DNA binding stronger on polystyrene MaxiSorp plates.

      Helpful?

  2. Bonjour,  Est ce que cette PLL est stérile ? Est-il possible de la filtrer ? Je vous remercie. Cordialement. Mme Boroni

    1 answer
    1. Solutions of this product are not sterile and contain a preservative, ProClin, to control contamination. In theory you could sterile filter P8920 through low protein binding filters, such as PVDF or PES. Sterile filtering will not remove the ProClin from P8920. Live cells should not be exposed to ProClin, as ProClin interrupts the Krebs cycle at 4 different sites. It would be best to purchase the powder as P1524 and then sterile filter after the solution has been prepared.

      Helpful?

  3. Hello! How many microscopy slides can I adhesive for every 100 milliliters of this solution( Poli-L- Lisine),? Gratitude!

    1 answer
    1. The number of slides that may be coated is dependent upon the surface area. The general use instructions recommend using 0.5 - 1.0 mL per 25 cm2 coating area - the 25 cm2 value is based on a T-25 tissue culture flask. A standard microscope slide has an approximate area of 1.3 cm2.

      Please see the link below to review the product datasheet which included a general protocol for coating: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/784/704/p6407pis.pdf

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. Is Product P8920, Poly-L-lysine solution, recommended for cell culture use?

    1 answer
    1. No, the product contain 0.01% thimerosal as a preservative. For cell culture use, product P4832 is recommended.  This solution has the same molecular weight as P8920 but contains no preservative.  P4832 is sterile filtered and is considered suitable for cell culture use.

      Helpful?

  6. What are the recommended uses for Product P8920, Poly-L-lysine solution?

    1 answer
    1. P8920 was developed for use in coating slides intended for use in either histology or immunohistochemistry.  Poly-L-lysine is preferred for uses such as silver staining procedures that require an alkaline pH.  Silane coated slides may be more effective is a low pH is required.

      Helpful?

Reviews

1 of 2 reviewers received a sample product or took part in a promotion

Active Filters

  1. Des Plaines, Illinois
    • Review 1
    • Votes 0
    5 out of 5 stars.

    stick with Poly-L-Lysine

    first purchased in 1993
    has been used regularly in our production ever since
    performs flawlessly

    Helpful?

  2. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service