Skip to Content
Merck
All Photos(8)

Documents

A2066

Sigma-Aldrich

Anti-Actin antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Actin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42 kDa

species reactivity

wide range, vertebrates, human, slime mold, amoeba, chicken

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25 using human or animal tissue sections
indirect immunofluorescence: 1:40 using chicken fibroblasts
western blot: 1:100 using chicken gizzard extract

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)
mouse ... Acta1(11459)
rat ... Acta1(29437)

General description

Actin, a highly conserved protein, is a major component of both the cytoskeletal and contractile structures in all cell types. It varies in amount, being related to the type of differentiation and to the functional state of cells and tissues. Actin can be found in two different forms of aggregation, the globular or the fibrillar state, and at least six distinct isoforms occur in vertebrates. The actins exhibit over 90% sequence homology, but each isoform has a unique NH2-terminal sequence.
The ACTA1 (actin, α 1) gene is mapped to human chromosome 1q42.13. The gene codes for actin 1, a thin filament component of skeletal muscle.

In immunoblotting, the product localizes actin in many species ranging from human skeletal muscle to amoeba. The product recognizes the 42 kDa actin band using immunoblotting techniques in human or animal tissue extracts.

Specificity

Anti-Actin specifically stains typical stress fibers in cultured chicken fibroblasts.

Immunogen

The C-terminal actin fragment attached to Multiple Antigen Peptide (MAP) backbone.

Application

May be used to study actin structure and function and to probe binding sites of actin-binding proteins.
Rabbit anti-actin antibody can be used for western blot assays. The antibody can also be used for immunohistochemistry (1:25, using human or animal tissue sections) and indirect immunofluorescence (1:40, using chicken fibroblasts) applications.
Used to study actin structure and function and to probe binding sites of actin-binding proteins. Protein lysates generated from embryonic mouse limb tissue was analyzed by western blot using actin as the loading control. Actin was detected using rabbit anti-actin at 1:8000. Actin detected by a rabbit anti-actin antibody was used as a loading control for MCF7 cell lysates. Actin was detected at 42 kDa.
Used to study actin structure and function and to probe binding sites of actin-binding proteins. Protein lysates generated from embryonic mouse limb tissue was analyzed by western blot using actin as the loading control. Actin was detected using rabbit anti-actin at 1:8000. Actin detected by a rabbit anti-actin antibody was used as a loading control for MCF7 cell lysates . Actin was detected at 42 kDa.

Biochem/physiol Actions

Actin is a cytoskeletal protein that regulates cell motility, secretion, phagocytosis and cytokinesis. The NH2-terminal of actin may function as an antigen. This terminal may also modulate actin interactions and may associate with proteins such as myosin. Actin protein is essential for the formation of mature platelets.
Mutations in ACTA1 (actin, α 1) is known to cause thin filament myopathy such as nemaline myopathy, congenital myopathy, actin myopathy, intranuclear rod myopathy, cap myopathy and core myopathy. Upregulation of the gene is observed in orthotopic liver transplantation without any earlier evidence of autoimmune liver disease. The encoded protein actin forms a network of filaments and is responsible for the shape of the cell. It provides cellular strength and promotes active movement of the cell.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytoskeletal regulation of platelet formation: Coordination of F-actin and microtubules
Poulter N S and Thomas S G
The International Journal of Biochemistry & Cell Biology, 66 (2015)
Prevalence and phenotypes of congenital myopathy due to α-actin 1 gene mutations.
Witting N, et al.
Muscle and Nerve, 53(3), 388-393 (2016)
Carl Robert Rankin et al.
Life sciences, 231, 116571-116571 (2019-06-18)
The role of long non-coding RNA's (lncRNA) in the biology of ulcerative colitis (UC) is not well understood. We have previously detected changes in lncRNA's associated with UC. This study aims to characterize one specific lncRNA, CDKN2B-AS1 whose expression was
Mutation-specific effects on thin filament length in thin filament myopathy.
Winter JM, et al.
Annals of Neurology, 79(6), 959-969 (2016)
Severe congenital actin related myopathy with myofibrillar myopathy features.
Selcen D.
Neuromuscular Disorders, 25(6), 488-492 (2015)

Articles

Loading controls in western blotting application.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service