Skip to Content
Merck
All Photos(4)

Documents

P5758

Sigma-Aldrich

Phenolphthalein monophosphate bis(cyclohexylammonium) salt

powder, ≥85% (TLC)

Synonym(s):

4-hydroxyphenyl 4-(3-oxo-1,3-dihydro-2-benzofuran-1-yl)phenyl hydrogen phosphate bis(cyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15O7P · 2C6H13N
CAS Number:
Molecular Weight:
596.65
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥85% (TLC)

form

powder

impurities

≤10% solvent

color

white to off-white

solubility

methanol: 10 mg/mL, clear to very slightly hazy, colorless

storage temp.

−20°C

SMILES string

NC1CCCCC1.NC2CCCCC2.Oc3ccc(cc3)C4(OC(=O)c5ccccc45)c6ccc(OP(O)(O)=O)cc6

InChI

1S/C20H15O7P.2C6H13N/c21-15-9-5-13(6-10-15)20(18-4-2-1-3-17(18)19(22)26-20)14-7-11-16(12-8-14)27-28(23,24)25;2*7-6-4-2-1-3-5-6/h1-12,21H,(H2,23,24,25);2*6H,1-5,7H2

InChI key

HANAZWFQWVNYKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenolphthalein monophosphate bis(cyclohexylammonium) salt has been used as a standard substrate in competitive enzyme-linked immunosorbent assay (ELISA) to determine the progesterone (P4) concentration in the whole milk samples.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ines Adriaens et al.
Theriogenology, 103, 44-51 (2017-08-06)
Reproductive performance is an important factor affecting the profitability of dairy farms. Optimal fertility results are often confined by the time-consuming nature of classical heat detection, the fact that high-producing dairy cows show estrous symptoms shorter and less clearly, and

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service