Skip to Content
Merck
All Photos(2)

Key Documents

S4762

Sigma-Aldrich

Streptavidin from Streptomyces avidinii

essentially salt-free, lyophilized powder, ≥13 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

form

essentially salt-free, lyophilized powder

Quality Level

specific activity

≥13 units/mg protein

mol wt

~60 kDa

capacity

4 mol/mol (Biotin)

application(s)

research pathology

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Streptavidin derives its name from its bacterial source Streptomyces avidinii and from the hen egg-white protein, avidin, which has high affinity to biotin. Its homologous core shares 33% sequence similarity with avidin, as well as sharing a common tetrameric structure. It is a crystalline tetrameric protein, with a molecular weight of 4*15000Da. It binds four molecules of biotin. Streptavidin lacks carbohydrate and sulfur-containing amino acids.
Streptavidin is a crystalline protein isolated from the bacterium Streptomyces avidinii and possesses biotin-binding ability. It has four binding sites for biotin like avidin and binds four molecules of biotin. Streptavidin is now considered as a new type of biotin-binding protein.

Application

Streptavidin from Streptomyces avidinii has been used:
  • in wire functionalization
  • in streptavidin capture enzyme-linked immunosorbent assay (ELISA)
  • in microbubble preparation

Preparation Note

Affinity purified.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid streptavidin-capture ELISA specific for the detection of antibodies to feline foamy virus.
Winkler IG et al
Journal of Immunological Methods, 207(1), 69-77 (1997)
Mapping of functional domains in the p22(phox) subunit of flavocytochrome b(559) participating in the assembly of the NADPH oxidase complex by "peptide walking".
Dahan I et al
The Journal of Biological Chemistry, 277(10), 8421-8432 (2002)
Viability of endothelial cells after ultrasound-mediated sonoporation: Influence of targeting, oscillation, and displacement of microbubbles
van R T, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 238, 197-211 (2016)
Isolation and properties of streptavidin.
E A Bayer et al.
Methods in enzymology, 184, 80-89 (1990-01-01)
Biotinylated CdSe/ZnSe nanocrystals for specific fluorescent labeling.
Charvet N et al
Journal of Materials Chemistry, 14.17 , 2638-2642 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service