Skip to Content
Merck
All Photos(2)

Key Documents

MAK159

Sigma-Aldrich

Mitochondrial Membrane Potential Kit

sufficient for 500 fluorometric tests (microplate readers)

Synonym(s):

JC-10 Assay, JC-10 Mitochondrial Membrane Potential Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 500 fluorometric tests (microplate readers)

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Mitochondria generate a potential across their membranes due to the activities of enzymes of the electron transport chain. During apoptosis, collapse of the mitochondrial membrane potential (MMP) coincides with the opening of the mitochondrial permeability transition pores, leading to the release of cytochrome c into the cytosol, which in turn triggers other downstream events in the apoptotic cascade.

Application

Mitochondrial Membrane Potential Kit has been used to measure mitochondrial membrane potential.

Suitability

This kit is suitable for the detection of Mitochondrial Membrane Potential in mammalian cells and for screening apoptosis inhibitors and activators using microplate readers.

Principle

This kit utilizes JC-10, a superior alternative to JC-1, for determining the loss of the MMP in cells. Although JC-1 is widely used in many labs, its poor water solubility often results in precipitation in aqueous buffers when used at higher concentrations. At higher concentrations, JC-10 exhibits greater aqueous solubility than JC-1. Similar to JC-1, JC-10 is a cationic, lipophilic dye that is concentrated and forms reversible red-fluorescent JC-10 aggregates (λex = 540/λem = 590 nm) in the mitochondria of cells with a polarized mitochondrial membrane. In apoptotic cells, MMP collapse results in the failure to retain JC-10 in the mitochondria and a return of the dye to its monomeric, green fluorescent form (λex = 490/λem = 525 nm). This kit can be used for monitoring apoptosis and for screening apoptosis inhibitors and activators.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Yang et al.
Biochemical and biophysical research communications, 517(1), 111-117 (2019-07-16)
Doxorubicin (DOX) is a potent anti-neoplastic agent with cumulative cardiotoxicity. DOX-induced cardiotoxicity has been shown to depend on the different dosing times. However, the basis for determining the dosing time to minimize DOX-induced cardiotoxicity and the underlying mechanisms remain incompletely
The mitochondrial membrane potential (deltapsi(m)) in apoptosis.
Jennifer D L, et al.
Apoptosis, 8, 116-128 (2003)
Hemlata et al.
ACS omega, 6(15), 10383-10395 (2021-06-01)
In recent decades, biodegradable polymeric nanoparticles have been used as a nanocarrier for the delivery of anticancer drugs. In the present study, we synthesize bovine serum albumin (BSA) nanospheres and evaluate their ability to incorporate a plant extract with anticancer
Giulia Allavena et al.
Cell death & disease, 9(7), 780-780 (2018-07-15)
The cellular recycling pathway of autophagy plays a fundamental role in adaptive responses to nutrient deprivation and other forms of stress under physiological and pathological conditions. However, autophagy can also be a double-edge sword during certain bacterial infections (such as
Rui Zhang et al.
Brain research, 1652, 135-143 (2016-10-04)
Abnormal gene expression, including mRNAs, and microRNAs (miRNA), have been identified in the development of Alzheimer's disease (AD). Although mitofusin2 (mfn2) has been found to be down-regulated in the neurons from hippocampus and cortex in AD patients, little is known

Articles

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Cellular apoptosis assays to detect programmed cell death using Annexin V, Caspase and TUNEL DNA fragmentation assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service