Skip to Content
Merck
All Photos(2)

Key Documents

MAK144

Sigma-Aldrich

Fluorometric Intracellular Ros Kit

sufficient for 200 fluorometric tests (orange)

Synonym(s):

Fluorometric ROS Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.25

usage

sufficient for 200 fluorometric tests (orange)

application(s)

cosmetics
food and beverages

detection method

fluorometric

relevant disease(s)

cancer; neurological disorders

storage temp.

−20°C

General description

Reactive oxygen species (ROS) are generated as a result of the reduction of oxygen during aerobic respiration and by various enzymatic systems within the cell. At physiological levels, ROS contribute to cell signaling and host defense. Increased ROS generation above the biological system′s detoxification capacity results in oxidative stress and cellular damage. The main damage to cells results from the ROS-induced alteration of macromolecules such as polyunsaturated fatty acids in membrane lipids, essential proteins, and DNA. ROS has been implicated in disease states, such as Alzheimer′s disease, Parkinson′s disease, cancer, and aging.

Application

Fluorometric intracellular reactive oxygen species (ROS) Kit has been used to evaluate the mitochondrial damage by mechanical ventilation (MV). It is also used to measure the production of intracellular ROS.

Suitability

This kit is suitable for the detection of intracellular ROS by fluorescence microplate reader, flow cytometer, or a fluorescent microscope with TRITC filter.

Principle

The Fluorometric Intracellular ROS Assay Kit provides a sensitive, one-step fluorometric assay to detect intracellular ROS (especially superoxide and hydroxyl radical) in live cells within 1 hour incubation. ROS react with a cell-permeable sensor, resulting in a fluorometric product (λex = 540/λem = 570 nm) proportional to the amount of ROS present. The assay can be performed in a convenient 96-well or 384-well microtiter-plate format.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidized low density lipoprotein (ox-LDL) binding to ox-LDL receptor-1 in endothelial cells induces the activation of NF-KB through an increased production of intracellular reactive oxygen species.
Cominacini L, et al.
The Journal of Biological Chemistry, 275(17), 12633-12638 (2000)
A mitochondrial paradigm of metabolic and degenerative diseases, aging, and cancer: a dawn for evolutionary medicine.
Wallace D C.
Annual Review of Genetics, 39, 359-407 (2005)
Bora Yim et al.
Investigative ophthalmology & visual science, 58(1), 362-371 (2017-01-25)
Silica nanoparticles (SiNPs) are promising carriers for ophthalmic drug delivery. In this study, we investigated the effect of various sizes of nonporous SiNPs on cultured human keratocytes. Three different sizes of SiNPs (50, 100, and 150 nm) were manufactured. Primarily
Neurodegenerative diseases and oxidative stress.
Barnham K J, et al.
Nature Reviews. Drug Discovery, 3(3), 205-205 (2004)
Tao Zheng et al.
Frontiers in pharmacology, 12, 669146-669146 (2021-05-25)
Although miR-327 had a protective effect on cardiomyocytes as described previously, the potential mechanism still needs further exploration. The aim of this study was to investigate the role and mechanism of miR-327 on oxidative stress in myocardial ischemia/reperfusion injury (MI/RI)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service