Skip to Content
Merck
All Photos(3)

Key Documents

M5546

Sigma-Aldrich

Anti-c-Myc Tag Antibody

mouse monoclonal, 9E10

Synonym(s):

Anti-c-Myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

9E10, monoclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:100 using c-myc tagged fusion protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

MYC (MYC proto-oncogene) is a transcriptional factor and oncoprotein. c-myc is a member of MYC gene family. It is located on human chromosome 8q24. c-Myc gene codes for basic helix-loop-helix/leucine zipper (bHLH/LZ) transcription factor.
Monoclonal Anti-c-myc (mouse IgG1 isotype) is derived from the 9E10 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from immunized BALB/c mice.
c-Myc is a transcription factor which is over-expressed in a wide range of cancers.

Specificity

Recognizes both N- terminal and C-terminal c-Myc tagged proteins.

Immunogen

synthetic peptide of the human p62c-Myc protein.

Application

Monoclonal anti-c-Myc antibody produced in mouse has been used for Western blotting and immunohistochemistry.
Monoclonal Anti-c-Myc antibody has also been used in:
  • the detection of horseradish peroxidase conjugate in phosphate buffered saline tween-20 (PBST) and 65-kDa mannoprotein (MP65) Dabs
  • immunoblotting
  • immunoprecipitation
  • biochemical and histological studies

Biochem/physiol Actions

The cellular myelocytomatosis (c-myc) oncogene plays a major role in cellular proliferation, differentiation and apoptosis. It acts as a transcriptional regulator of gene expression. MYC (MYC proto-oncogene) may be essential for cancer initiation, promotion and therapy resistance. Overexpression of MYC in DLBCL (diffuse large B-cell lymphoma) results in the poor outcome and invasive treatment, when medicated with rituximab plus cyclophosphamide, doxorubicin, vincristine and prednisone (R-CHOP).
c-Myc is a regulator of the cell cycle and aids in the growth of tumors in the liver. Translocations in the gene encoding it have been shown to be linked to Burkitt lymphoma. It has been studied that c-Myc targets many protein-coding genes which enhances the oncogenic properties of c-Myc.

Physical form

The product is provided as ascites fluid with 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for a maximum of one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human domain antibodies against virulence traits of Candida albicans inhibit fungus adherence to vaginal epithelium and protect against experimental vaginal candidiasis.
De Bernardis F, et al.
The Journal of Infectious Diseases, 195(1), 149-157 (2007)
MAPK signal pathways in the regulation of cell proliferation in mammalian cells.
Zhang W and Liu HT
Cell Research, 12(1), 9-9 (2002)
Basis for the isoform-specific interaction of myosin phosphatase subunits protein phosphatase 1c β and myosin phosphatase targeting subunit 1.
Scotto-Lavino E, et al.
The Journal of Biological Chemistry, 285(9), 6419-6424 (2010)
Regulation of Apobec3F and human immunodeficiency virus type 1 Vif by Vif-Cul5-ElonB/C E3 ubiquitin ligase.
Liu B, et al.
Journal of Virology, 79(15), 9579-9587 (2005)
c-Myc is essential for vasculogenesis and angiogenesis during development and tumor progression.
Baudino TA, et al.
Genes & Development, 16(19), 2530-2543 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service