Skip to Content
Merck
All Photos(1)

Key Documents

ROAMYC

Roche

Anti-c-myc

from mouse IgG1κ

Synonym(s):

anti-myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

Assay

90% (SDS-PAGE)

form

frozen liquid (11667203001)
lyophilized (11667149001)

packaging

pkg of 200 μg (11667149001)
pkg of 5 mg (11667203001 [1 ml])

manufacturer/tradename

Roche

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

EQKLISEEDL

storage temp.

−20°C

General description

Anti-c-myc is a monoclonal antibody to c-myc peptide (clone 9E10). Anti-c-myc recognizes the 9E10 epitope sequence (EQKLISEEDL), derived from the human c-myc protein. The monoclonal antibody against the c-myc epitope is well characterized and does not cross-react with other cellular proteins. The antibody recognizes its antigenic determinant even when the c-myc peptide epitope is introduced into unrelated recombinant proteins by a technique known as epitope tagging.
The cellular myelocytomatosis (c-myc) is the cellular homologue of the v-myc gene originally isolated from an avian myelocytomatosis virus. The gene encoding it is mapped to human chromosome 8q24. It c-myc is a member of MYC gene family. c-Myc gene codes for basic helix-loop-helix/leucine zipper (bHLH/LZ) transcription factor that regulates the G1-S cell cycle transition.


Contents:
  • 11 667 149 001: lyophilizate, stabilized
  • 11 667 203 001: frozen solution, (5 mg/ml)

Specificity

Anti-c-myc recognizes the amino acid sequence EQKLISEEDL, which was derived from the human c-myc protein. The antibody recognizes its antigenic determinant even when the c-myc peptide epitope is introduced into unrelated recombinant proteins either N-terminal, C-terminal or internal. The monoclonal antibody against the c-myc epitope is well characterized and does not crossreact with other cellular proteins.

Immunogen

Amino acid sequence EQKLISEEDL was derived from the human c-myc protein.

Application

Use Anti-c-myc for the detection of native human c-myc protein and recombinant epitope-tagged proteins that contain the c-myc epitope using:
  • Dot blots
  • Immunocytochemistry
  • Immunoprecipitation
  • Western blots
  • Immunohistochemistry.

Biochem/physiol Actions

The cellular myelocytomatosis (c-myc) oncogene has a crucial role in cellular proliferation, differentiation, apoptosis and acts as transcriptional regulator of gene expression. c-myc expression is essential and sufficient to assist most of the cells to enter DNA synthetic (S) phase of the cell cycle. The encoded protein plays a crucial role in vasculogenesis and angiogenesis during cancer development and progression. c-myc interacts with its binding partner Max and activates the transcription of growth promoting genes such as cyclin D2 and ornithine decarboxylase. It also represses the transcription of multiple genes, especially p21 and p27, by binding to the transcription initiator element (Inr) in a complex with Max and either Sp1 (specificity protein 1) or Miz1 (Myc-interacting zinc finger protein 1). Overexpression of MYC in DLBCL (diffuse large B-cell lymphoma) results in poor outcome and invasive treatment when medicated with rituximab plus cyclophosphamide, doxorubicin, vincristine and prednisone (R-CHOP).

Quality

Anti-c-myc antibody is tested for functionality and purity relative to a reference standard to confirm the quality of each new reagent lot.

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate.

The following concentrations should be taken as a guideline:
  • Western blot: 1 to 10 μg/ml


Working solution: Tris-buffered saline containing 0.1% Tween 20.

Reconstitution

Add 500 μl PBS to a final concentration of 400 μg/ml.

Analysis Note

The monoclonal antibody against the c-myc epitope is well characterized and does not crossreact with other cellular proteins. No cross reaction with murine c-myc.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ines B Folger et al.
Nature chemical biology, 20(6), 761-769 (2024-02-03)
Engineered biosynthetic assembly lines could revolutionize the sustainable production of bioactive natural product analogs. Although yeast display is a proven, powerful tool for altering the substrate specificity of gatekeeper adenylation domains in nonribosomal peptide synthetases (NRPSs), comparable strategies for other
Apoptotic signaling by c-MYC.
Hoffman B and Liebermann D A
Oncogene, 27(50), 6462-6472 (2008)
Over-expression of the c-myc proto-oncogene in colorectal carcinoma.
Smith DR, et al.
British Journal of Cancer (1993)
R Dalla-Favera et al.
Proceedings of the National Academy of Sciences of the United States of America, 79(24), 7824-7827 (1982-12-01)
Human sequences related to the transforming gene (v-myc) of avian myelocytomatosis virus (MC29) are represented by at least one gene and several related sequences that may represent pseudogenes. By using a DNA probe that is specific for the complete gene
MAPK signal pathways in the regulation of cell proliferation in mammalian cells.
Cell Research, 12(1), 9-18 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service