Skip to Content
Merck
All Photos(1)

Documents

M2449

Sigma-Aldrich

MB-3

≥95% (HPLC)

Synonym(s):

γ-butyrolactone, Butyrolactone 3, (2R,3S)-rel-4-Methylene-5-oxo-2-propyltetrahydrofuran-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O4
Molecular Weight:
184.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: ≥10 mg/mL
DMSO: ≥20 mg/mL

storage temp.

−20°C

SMILES string

CCC[C@H]1OC(=O)C(=C)[C@@H]1C(O)=O

InChI

1S/C9H12O4/c1-3-4-6-7(8(10)11)5(2)9(12)13-6/h6-7H,2-4H2,1H3,(H,10,11)/t6-,7+/m1/s1

InChI key

SRQUTZJZABSZRQ-RQJHMYQMSA-N

Application

MB-3 has been used as a general control non-repressible 5 (GCN5) inhibitor:
  • to study its effects on lamin A/C-associated pY19-Cav-2-mediated histone H3 acetylations
  • to analyze its effects on the growth of Arabidopsis thaliana seedlings
  • to study its effects on craniofacial chondrocyte maturation

Biochem/physiol Actions

MB-3 can modify the gene expression in murine embryonic stem cells as well as yeast. It hinders the acetyltransferase activity of general control non-repressible 5 (GCN5) by binding to acetyl CoA pocket present in GCN5.
MB-3 is a Gcn5 HAT (Histone Acetyltransferase) inhibitor; anti-inflammatory, anti-carcinogenic.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Xu et al.
Scientific reports, 10(1), 1817-1817 (2020-02-06)
Mesenchymal stem cells (MSCs) specifically differentiate into cardiomyocytes as a potential way to reverse myocardial injury diseases, and uncovering this differentiation mechanism is immensely important. We have previously shown that histone acetylation/methylation and DNA methylation are involved in MSC differentiation
Naomi Moris et al.
Stem cells (Dayton, Ohio), 36(12), 1828-1838 (2018-10-03)
Cell fate transitions in mammalian stem cell systems have often been associated with transcriptional heterogeneity; however, existing data have failed to establish a functional or mechanistic link between the two phenomena. Experiments in unicellular organisms support the notion that transcriptional
Hong-Bin Cai et al.
Journal of Alzheimer's disease : JAD, 77(1), 75-84 (2020-04-21)
The connection between diabetes and Alzheimer's disease (AD) is not fully determined. Hyperphosphorylation of tau protein is mediated by binding and stabilization of truncated p25 with cyclin-dependent kinase-5 (CDK5) in AD. We recently showed that diabetes-associated hyperglycemia increased the CDK5
Felipe Aquea et al.
Biochemical and biophysical research communications, 483(1), 664-668 (2016-12-21)
Chemical inhibition of chromatin regulators provides an effective approach to investigate the roles of chromatin modifications in plant and animals. In this work, chemical inhibition of the Arabidopsis histone acetyltransferase activity by γ-butyrolactone (MB-3), the inhibitor of the catalytic activity
Anissa Guillemin et al.
PloS one, 14(11), e0225166-e0225166 (2019-11-22)
To better understand the mechanisms behind cells decision-making to differentiate, we assessed the influence of stochastic gene expression (SGE) modulation on the erythroid differentiation process. It has been suggested that stochastic gene expression has a role in cell fate decision-making

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service