Skip to Content
Merck
All Photos(1)

Documents

N1537

Sigma-Aldrich

NU7026

≥98% (HPLC), solid

Synonym(s):

2-(Morpholin-4-yl)-benzo[h]chromen-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15NO3
CAS Number:
Molecular Weight:
281.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

tan

solubility

DMSO: soluble 3 mg/mL at 60 °C
H2O: insoluble

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O=C1C=C(Oc2c1ccc3ccccc23)N4CCOCC4

InChI

1S/C17H15NO3/c19-15-11-16(18-7-9-20-10-8-18)21-17-13-4-2-1-3-12(13)5-6-14(15)17/h1-6,11H,7-10H2

InChI key

KKTZALUTXUZPSN-UHFFFAOYSA-N

Application

NU7026 has been used:
  • as a DNA-dependent protein kinase (DNA-PK) inhibitor, to pre-treat colon cancer cell
  • in kinase inhibitor experiments for treating zygotes with visible pronuclei post 20 h-human chorionic gonadotropin (hCG)
  • to determine its influence on cytotoxicity of dibenzo[def,p]chrysene on HepG2 cells

Potent, ATP-competitive. IC50 = 0.23 μM. Selectivity over other PI3K-related kinases (IC50 = 13 μM for PI3K and >100 μM for ATM and ATR).

Biochem/physiol Actions

Cell permeable, small molecule benzochromenone inhibitor of DNA-PK (DNA dependent protein kinase).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Yasuda et al.
Genes to cells : devoted to molecular & cellular mechanisms, 26(5), 328-335 (2021-02-25)
SIRT2 and SIRT3 protein deacetylases maintain genome integrity and stability. However, their mechanisms for maintaining the genome remain unclear. To examine the roles of SIRT2 and SIRT3 in DSB repair, I-SceI-based GFP reporter assays for HR, single-strand annealing (SSA) and
Rachel E Doherty et al.
Cancers, 11(9) (2019-09-05)
Uveal melanoma (UM) is the most common primary intraocular tumour in adults, with a mean survival of six months following metastasis. The survival rates have not improved in over 30 years. This study has shown that sister chromatid exchange (SCE)
The influence of ATM, ATR, DNA-PK inhibitors on the cytotoxic and genotoxic effects of dibenzo [def, p] chrysene on human hepatocellular cancer cell line HepG2
Spryszynska S, et al.
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 791, 12-24 (2015)
Soichiro Shiga et al.
Biochemical and biophysical research communications, 521(3), 668-673 (2019-11-05)
Presence of unperfused regions containing cells under hypoxia and nutrient starvation; contributes to radioresistance in solid human tumors. We have previously reported that cultured cells; under nutrient starvation show resistance to ionizing radiation compare with cells under normal; condition, and
Chk1 and DNA-PK mediate TPEN-induced DNA damage in a ROS dependent manner in human colon cancer cells
Rahal ON, et al.
Cancer Biology & Therapy, 17(11), 1139-1148 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service