Skip to Content
Merck
All Photos(2)

Key Documents

L7875

Sigma-Aldrich

DL-Leucine

≥99% (HPLC)

Synonym(s):

(±)-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
636005
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

DL-Leucine, ≥99% (HPLC)

Assay

≥99% (HPLC)

form

powder

color

white

mp

293-296 °C (subl.) (lit.)

solubility

1 M HCl: soluble

application(s)

detection
peptide synthesis

SMILES string

CC(C)CC(N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)

InChI key

ROHFNLRQFUQHCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Leucine is a racemic mixture of the D- and L- enantiomers of Leucine. DL-Leucine is used to form benzimidazole and pyrimidine hydroxy azo dyes with various transition metals. DL-Leucine may be used for the evaluation of chiral amino acid separation techniques.

DL-Leucine is used for the following applications:
  • Used as a standard for the measurement of free amino acids
  • Used in the process of tritiated leucine uptake
  • Used in solution preparation (various organics were used in different combinations and concentrations to model complex surface tension effects, DL-Leucine is one of them). It is chosen due to its presence in atmospheric aerosols and ice nucleation activity as well as for its surfactant character

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Chang et al.
British journal of haematology, 111(1), 146-156 (2000-11-25)
Transfusion-associated graft-versus-host disease (TA-GVHD) is usually a fatal outcome of blood transfusion therapy, caused by viable leucocytes contained in the donor blood. Most cases of TA-GVHD occur when less than 4-d-old blood is transfused. We therefore examined the molecular changes
Modeling the surface tension of complex, reactive organic-inorganic mixtures.
A. N. Schwier et al.
Atmospheric Chemistry and Physics, 13, 10721-10732 (2013)
L Sallami et al.
Journal of dairy science, 87(6), 1585-1594 (2004-09-30)
The effect of incorporating a highly autolytic strain (Lactobacillus delbrueckii subsp. bulgaricus UL12) a proteolytic strain (Lactobacillus casei subsp. casei L2A), or a nisin Z-producing strain (Lactococcus lactis, subsp. lactis biovar diacetylactis UL719) into Cheddar cheese starter culture (Lactococcus lactis
Satinder K Singh et al.
Nature, 448(7156), 952-956 (2007-08-10)
Sodium-coupled transporters are ubiquitous pumps that harness pre-existing sodium gradients to catalyse the thermodynamically unfavourable uptake of essential nutrients, neurotransmitters and inorganic ions across the lipid bilayer. Dysfunction of these integral membrane proteins has been implicated in glucose/galactose malabsorption, congenital
Farid Ichou et al.
Journal of mass spectrometry : JMS, 49(6), 498-508 (2014-06-11)
Reproducibility among different types of excitation modes is a major bottleneck in the field of tandem mass spectrometry library development in metabolomics. In this study, we specifically evaluated the influence of collision voltage and activation time parameters on tandem mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service