Skip to Content
Merck
All Photos(1)

Key Documents

I0460000

Isoleucine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Isoleucine, (2S,3S)-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721792
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

leucine

manufacturer/tradename

EDQM

mp

288 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoleucine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jirí Svoboda et al.
Phytochemistry, 71(13), 1445-1449 (2010-06-24)
Our understanding of plant defensive mechanisms against herbivore and pathogen attack has significantly increased over the past decade. The complex cascade of defensive events is initiated and controlled by a network of interacting plant hormones. Especially, the conjugate of jasmonate
Anna L Vestergaard et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(14), E1334-E1343 (2014-04-08)
P4-ATPases (flippases) translocate specific phospholipids such as phosphatidylserine from the exoplasmic leaflet of the cell membrane to the cytosolic leaflet, upholding an essential membrane asymmetry. The mechanism of flipping this giant substrate has remained an enigma. We have investigated the
Vijay Joshi et al.
Amino acids, 39(4), 933-947 (2010-02-27)
Pathways regulating threonine, methionine and isoleucine metabolism are very efficiently interconnected in plants. As both threonine and methionine serve as substrates for isoleucine synthesis, their synthesis and catabolism under different developmental and environmental conditions also influence isoleucine availability. Together, methionine
Song-Yu Yang et al.
Molecular genetics and metabolism, 92(1-2), 36-42 (2007-07-10)
The HSD17B10 gene maps on chromosome Xp11.2, a region highly associated with X-linked mental retardation. This gene encodes HSD10, a mitochondrial multifunctional enzyme that plays a significant part in the metabolism of neuroactive steroids and the degradation of isoleucine. The
O Søvik
Journal of inherited metabolic disease, 16(1), 46-54 (1993-01-01)
A review is presented of 22 published cases of verified or probable mitochondrial 2-methylacetoacetyl-CoA thiolase deficiency, a disorder of isoleucine and ketone body metabolism. The clinical expression, characterized by ketoacidosis, vomiting and lethargy, is highly variable. Typical age of onset

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service