Skip to Content
Merck
All Photos(3)

Key Documents

147966

Sigma-Aldrich

DL-Phenylalanine

99%, for peptide synthesis, ReagentPlus®

Synonym(s):

(±)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
165.19
Beilstein:
3198807
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

DL-Phenylalanine, ReagentPlus®, 99%

product line

ReagentPlus®

Assay

99%

form

powder, crystals or granules

reaction suitability

reaction type: solution phase peptide synthesis

mp

266-267 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylalanine is an amino acid. It participates in the preparation of ternary piroxicam complexes of Fe(II), Fe(III), Co(II), Ni(II), Cu(II) and Zn(II). Characterization of these complexes by elemental analyses, molar conductance, IR, UV-Vis, magnetic moment, diffuse reflectance and X-ray powder diffraction methods has been conducted. Crystalline DL-phenylalanine was prepared by its reduction in silica gel by solubility method.

Application

DL-Phenylalanine may be used in the synthesis of ternary piroxicam (Pir; 4-hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide) complexes of Fe(II), Fe(III), Co(II), Ni(II), Cu(II) and Zn(II).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

XRD, thermal and FTIR studies on gel grown DL-Phenylalanine crystals.
Ramachandran E and Natarajan S.
Crystal Research and Technology, 42(6), 617-620 (2007)
Gehad G Mohamed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(13), 3141-3154 (2004-10-13)
The ternary piroxicam (Pir; 4-hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide) complexes of Fe(II), Fe(III), Co(II), Ni(II), Cu(II) and Zn(II) with various amino acids (AA) such as glycine (Gly) or DL-phenylalanine (PhA) were prepared and characterized by elemental analyses, molar conductance, IR, UV-Vis, magnetic moment
Katarzyna M Romek et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8296-8301 (2015-06-25)
Tramadol, previously only known as a synthetic analgesic, has now been found in the bark and wood of roots of the African medicinal tree Nauclea latifolia. At present, no direct evidence is available as to the biosynthetic pathway of its
Raja R Pillai et al.
The Journal of nutrition, 145(5), 954-959 (2015-03-13)
Lysine requirements of well-nourished children from developing regions have been found to be similar to those of children from developed regions (33.5 mg · kg⁻¹ · d⁻¹). However, intestinal parasites have been shown to increase lysine requirements in undernourished adults, and it is not
Sohei Nakayama et al.
Cancer research, 74(20), 5891-5902 (2014-08-29)
The discovery of somatic mutations in EGFR and development of EGFR tyrosine kinase inhibitors (TKI) have revolutionized treatment for lung cancer. However, resistance to TKIs emerges in almost all patients and currently no effective treatment is available. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service