Skip to Content
Merck
All Photos(1)

Documents

L6007

Sigma-Aldrich

Leucine Aminopeptidase, microsomal from porcine kidney

Type VI-S, lyophilized powder, ≥12 units/mg protein (biuret)

Synonym(s):

Aminopeptidase M, Aminopeptidase N, Cathepsin III, LAP, Leucinamide aminopeptidase, Leucinaminopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Porcine kidney

Quality Level

type

Type VI-S

form

lyophilized powder

specific activity

≥12 units/mg protein (biuret)

mol wt

320 kDa

composition

Protein, 30-70%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Leucine Aminopeptidase (LAP) from porcine is a 320-kDa exopeptidase present in cytosol. It contains zinc in its structure and belongs to the M1 and M17 peptidase families. LAP display bi-lobed structure and has the catalytic domain at the C-terminus and a variable N-terminal domain.

Application

Leucine Aminopeptidase, microsomal from porcine kidney has been used in aminopeptidase N (APN) inhibitor screening assay in adenocarcinoma cell line ES-2 and in the inhibition kinetic studies with APN inhibitor.

Biochem/physiol Actions

Leucine Aminopeptidase (LAP) catalyzes protein hydrolysis at the leucine residue at N-terminal. An altered level of LAP is implicated in tumor proliferation and progression. LAP is a potential biomarker for ovarian epithelial cancer therapy. Porcine LAP hydrolyzes glycine and arginine substrates.

Unit Definition

One unit will hydrolyze 1.0 μmole of L-leucine-p-nitroanilide to L-leucine and p-nitroaniline per min at pH 7.2 at 37 °C. (At 25 °C, approx. 40% of the activity at 37 °C is obtained.) The activity obtained using L-leucine p-nitroanilide as substrate is 2-5 times that obtained with L-leucinamide as substrate.

Physical form

Contains primarily phosphate buffer salts, pH 7.0

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel and highly sensitive fluorescent assay for leucine aminopeptidases
Huang H, et al.
Analytical biochemistry, 391(1), 11-16 (2009)
Leucine aminopeptidases: diversity in structure and function
Matsui M, et al.
Biological Chemistry, 387(12), 1535-1544 (2006)
The effect of different species Aminopeptidase N Structure on the activity screening of Aminopeptidase N Inhibitor
Wang X, et al.
Biological & Pharmaceutical Bulletin, 33(10), 1658-1665 (2010)
Refolding of hexameric porcine leucine aminopeptidase using a cationic detergent and dextrin-10 as artificial chaperones
Laslo AC, et al.
Journal of Biotechnology, 140(3-4), 162-168 (2009)
Activity screening and structure-activity relationship of the hit compounds targeting APN/CD13
Wang X, et al.
Fundamental & Clinical Pharmacology, 25(2), 217-228 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service