Skip to Content
Merck
All Photos(1)

Key Documents

PRON-RO

Roche

Pronase

from Streptomyces griseus

Synonym(s):

non-specific protease

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

Streptomyces griseus

Quality Level

sterility

non-sterile

form

lyophilized powder

packaging

pkg of 1 g (10165921001)
pkg of 5 g (11459643001)

manufacturer/tradename

Roche

concentration

0.5-2 mg/mL

parameter

35-40 °C optimum reaction temp.

technique(s)

DNA extraction: suitable

color

white

optimum pH

7.0-8.0

pH range

6.0-9.0

solubility

water: soluble 10-20 g/L

suitability

suitable for molecular biology

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

foreign activity

Deoxyribonuclease <0.002 U/mg ( based on lyoph.)
Ribonuclease <0.002 U/mg ( based on lyoph.)

storage temp.

2-8°C

Gene Information

Streptomyces griseus ... SGR_RS27460(6214424)

Related Categories

General description

at 40°C with casein as the substrate, pH 7.5, equivalent to approximately 1270 PU/mg or approximately 25 PUK/mg.

Specificity

Pronase has a broad specificity, breaking down virtually all proteins into their individual amino acids; resolves carboxylic acids and alcohols.

Application

Use Pronase to completely hydrolyze proteins in research applications. Pronase is used for the degradation of proteins during the isolation of DNA and RNA, such as in the extraction of phage DNA or the isolation of plasmid DNA. It is not necessary to let pronase self-digest prior to use. It is also used in histochemistry and cell culture for tissue dissociation in conjunction with collagenase and trypsin,[1][2][3] and for the production of glycopeptides from purified glycoproteins. It has been used for the dechorionation of Zebrafish embryos.[4]

Biochem/physiol Actions

Pronase has wide-ranging specificity. Therefore, it is commonly used when extensive or complete degradation of proteins is required. Its applications include protein analysis in cell organelles, removal of proteins during DNA and RNA isolation, and the production of protein hydrolysates for amino acid analysis.[5] In addition, Pronase surpasses trypsin in its ability to disperse fibroblastic cell lines, excelling in both monolayer detachment and the creation of single-cell suspensions.[6]

Unit Definition

A unit of non-specific protease activity increases the rate of release of folin-positive amino acids and peptides from casein, at +40 °C and pH 7.5. For one unit (U) 1 μmol/min tyrosine is released, and for the unit "PU", the value is 1 μg/minute (1 U = 181 PU); for the unit "PUK", it is 0.1/minute (measured is the change in absorbance of molybdenum blue, formed by reaction with the folin ′s reagent under conditions such that 1 PUK = 50 PU).

Preparation Note

Stabilizers: Protective effect of calcium ions

The preparation contains 20% calcium acetate for stability. It is free of starch as per the current Quality Control Procedures.The activity of a diluted solution containing 0.01–0.1 M calcium was stable over 24 hours at neutral pH at 4 to 8 °C. Pronase is also protected from heat inactivation by low levels of calcium.
Working concentration: 0.5 to 2 mg/ml
Working solution: Solvent is recommended in distilled water.
Stock solution is prepared by adding pronase powder to distilled water (10 to 20 mg/ml).
Storage conditions (working solution): -15 to -25 °C

Reconstitution

The lyophilizate is soluble in water (10 to 20 mg/ml).

Storage and Stability

Store dry

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changhe Hou et al.
Molecular medicine reports, 12(4), 4877-4886 (2015-07-03)
The aim of the present study was to investigate the presence and biological function of microRNA-92a (miR-92a) in chondrogenesis and cartilage degeneration. Human adipose‑derived mesenchymal stem cells (hADSCs) in micromass and chondrocyte‑like ATDC5 cells were induced to chondrogenesis, and primary
Yue-Hong Chen et al.
Arthritis research & therapy, 24(1), 27-27 (2022-01-21)
Penfluridol, isolated from an FDA-approved small-molecule drug library as an inhibitor of tumor necrosis factor α (TNFα)-stimulated NF-κB activation, is clinically used to treat chronic schizophrenia and related disorders. This study is aimed to investigate the therapeutic effect of penfluridol
Pronase
Foley JF and Aftonomos BTh
Tissue Culture and Research, 185-188 (1973)
Chen Chen et al.
Journal of neuroinflammation, 19(1), 315-315 (2022-12-29)
Dysregulated activation of the inflammasome is involved in various human diseases including acute cerebral ischemia, multiple sclerosis and sepsis. Though many inflammasome inhibitors targeting NOD-like receptor protein 3 (NLRP3) have been designed and developed, none of the inhibitors are clinically available. Growing evidence
Tsung-Jung Ho et al.
Pharmaceuticals (Basel, Switzerland), 16(1) (2023-01-22)
Catechu is a dried decoction from twigs with the leaves of Uncaria gambir. Its antioxidant, anti-inflammatory, and antimicrobial activities have been previously reported because of its high catechin and epicatechin content (>21%). It is also one of the components used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service