Skip to Content
Merck
All Photos(2)

Key Documents

A6671

Sigma-Aldrich

Actinonin

≥98% (TLC), powder, leucine aminopeptidase inhibitor

Synonym(s):

3-[[1-[(2-(Hydroxymethyl)-1-pyrrolidinyl)carbonyl]-2-methylpropyl]carbamoyl]octanohydroxamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H35N3O5
CAS Number:
Molecular Weight:
385.50
Beilstein:
1555250
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Actinonin,

biological source

synthetic (organic)

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 50 mg/mL, clear, colorless

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

enzyme | inhibits

storage temp.

−20°C

SMILES string

CCCCCC(CC(=O)NO)C(=O)NC(C(C)C)C(=O)N1CCCC1CO

InChI

1S/C19H35N3O5/c1-4-5-6-8-14(11-16(24)21-27)18(25)20-17(13(2)3)19(26)22-10-7-9-15(22)12-23/h13-15,17,23,27H,4-12H2,1-3H3,(H,20,25)(H,21,24)

InChI key

XJLATMLVMSFZBN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Application

Actinonin has been used as a control to inhibit meprin-β activity in C57BL/6 mice. It has also been used to study the high-affinity interaction of EcPDF with actinonin by 15N NMR spectroscopy and isothermal titration.

Biochem/physiol Actions

Actinonin has inhibitory action against peptide deformylase (PDF). It is effective against Gram-positive and fastidious Gram-negative microorganisms.
Actinonin is an inhibitor of leucine aminopeptidase. Actinonin has also been used to improve antibacterial activity and antiobesity therapeutics.

Features and Benefits

This compound is featured on the Neuropeptidases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Actinonin, a naturally occurring antibacterial agent, is a potent deformylase inhibitor
Chen DZ, et al.
Biochemistry, 39(6), 1256-1262 (2000)
Diana Mader et al.
Microbes and infection, 12(5), 415-419 (2010-02-17)
The biosynthesis of proteins with N-terminal formylated methionine residues and subsequent protein deformylation are unique and invariant bacterial processes. They are exploited by the capacity of the human innate immune system to sense formylated peptides (FPs) and targeted by the
Sonia Fieulaine et al.
PLoS biology, 9(5), e1001066-e1001066 (2011-06-02)
For several decades, molecular recognition has been considered one of the most fundamental processes in biochemistry. For enzymes, substrate binding is often coupled to conformational changes that alter the local environment of the active site to align the reactive groups
Ligand-induced changes in the structure and dynamics of Escherichia coli peptide deformylase
Amero CD, et al.
Biochemistry, 48(32), 7595-7607 (2009)
Jennifer Hamner et al.
Scientific reports, 10(1), 6358-6358 (2020-04-15)
Vaginal delivery with obstetrical trauma is a risk factor for pelvic organ prolapse later in life. Loss of fibulin-5 (FBLN5), an elastogenesis-promoting cellular matrix protein, results in prolapse in mice. Here, we evaluated effects of pregnancy, parturition, and obstetrical injury

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service