L2752
Leu-Leu
>98% (TLC), suitable for ligand binding assays
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H24N2O3
CAS Number:
Molecular Weight:
244.33
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
Leu-Leu,
Assay
>98% (TLC)
form
powder
technique(s)
ligand binding assay: suitable
color
white to off-white
storage temp.
−20°C
SMILES string
CC(C)C[C@H](N)C(=O)N[C@@H](CC(C)C)C(O)=O
InChI
1S/C12H24N2O3/c1-7(2)5-9(13)11(15)14-10(12(16)17)6-8(3)4/h7-10H,5-6,13H2,1-4H3,(H,14,15)(H,16,17)/t9-,10-/m0/s1
InChI key
LCPYQJIKPJDLLB-UWVGGRQHSA-N
Related Categories
Amino Acid Sequence
Leu-Leu
Biochem/physiol Actions
Leucylleucine (Leu-Leu) may be used to study the functionality of dileucine motifs such as the motif responsible for internalization and targeting of vesicular acetylcholine transporter and clathrin adaptors AP-1 and AP-2.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Guoqing Wang et al.
Cellular and molecular neurobiology, 41(1), 163-171 (2020-04-23)
Traditionally, hydrogen peroxide (H2O2) was formed from cellular oxidative metabolism and often viewed as toxic waste. In fact, H2O2 was a benefit messenger for neuron-glia signaling and synaptic transmission. Thus, H2O2 was a double-edged sword and neuroprotection vs. neurotoxicity produced
Lingling Zhang et al.
Oncogene, 39(49), 7196-7208 (2020-10-11)
Metastasis is responsible for the death of most breast cancer patients. Robo1 has been implicated as a tumor suppressor for various cancers including breast cancer. However, it is not well understood how Robo1 expression is regulated during tumorigenesis. In this
Yanzhen Zhou et al.
Journal of neuroinflammation, 15(1), 161-161 (2018-05-29)
Microglia-mediated neuroinflammation is recognized to mainly contribute to the pathogenesis of Parkinson's disease (PD). Tetrahydroxystilbene glucoside (TSG) has been proved to be beneficial for health with a great number of pharmacological properties. We examined the effects of TSG against dopamine
A van Boven et al.
Biochimie, 70(4), 535-542 (1988-04-01)
Different strains of Lactococcus lactis ssp. cremoris hydrolyze peptides at different rates while the cell-free extracts of these strains all show the same or much higher rates of hydrolysis. These observations indicate that the uptake of peptides is the rate-limiting
S Martinez-Arca et al.
Journal of cell science, 113 ( Pt 10), 1705-1715 (2000-04-19)
The mechanisms by which the insulin-sensitive glucose transporter, GLUT4, is targeted and retained in a storage compartment near to the Golgi complex are poorly understood. Here we report that removal of the carboxyl-terminal acidic Pro(505)AspGluAsnAsp(509) sequence prevents the storage of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service