Skip to Content
Merck
All Photos(2)

Key Documents

P6258

Sigma-Aldrich

Phe-Pro

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O3
CAS Number:
Molecular Weight:
262.30
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

O=C(N1CCC[C@H]1C(O)=O)[C@@H](N)CC2=CC=CC=C2

InChI

1S/C14H18N2O3/c15-11(9-10-5-2-1-3-6-10)13(17)16-8-4-7-12(16)14(18)19/h1-3,5-6,11-12H,4,7-9,15H2,(H,18,19)

InChI key

WEQJQNWXCSUVMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Trapp et al.
Electrophoresis, 25(2), 318-323 (2004-01-27)
Dynamic capillary electrophoresis (DCE) and computer simulation of the elution profiles with the stochastic model has been applied to determine the isomerization barriers of the angiotensin converting enzyme inhibitor enalaprilat. The separation of the rotational cis-trans isomeric drug has been
Enrique Martínez-Carranza et al.
Extremophiles : life under extreme conditions, 22(1), 73-85 (2017-11-13)
The Cuatro Ciénegas Basin (CCB) within the Chihuahuan Desert in México is an extremely oligotrophic oasis with negligible phosphorous levels, described as a hot spot of biodiversity, not only in stromatolites and microbial mats, but also in living forms in
R Gurunath et al.
Biochemical and biophysical research communications, 202(1), 241-245 (1994-07-15)
Non protein amino acids with strong secondary structure preferences are potentially useful in peptide design. alpha-Aminoisobutyric acid (Aib) is a powerful 'stereochemical director' of polypeptide chain folding, stabilizing helical conformations in diverse oligopeptide sequences. In an approach to the de
Wooseong Lee et al.
Nature communications, 9(1), 1606-1606 (2018-04-25)
The recognition of pathogen-derived ligands by pattern recognition receptors activates the innate immune response, but the potential interaction of quorum-sensing (QS) signaling molecules with host anti-viral defenses remains largely unknown. Here we show that the Vibrio vulnificus QS molecule cyclo(Phe-Pro)
G Schoetz et al.
Electrophoresis, 22(12), 2409-2415 (2001-08-25)
Dynamic capillary electrophoresis (DCE) and computer simulation of the elution profiles with the theoretical plate and the stochastic model has been applied to determine the isomerization barriers of the three dipeptides L-alanyl-L-proline, L-leucyl-L-proline, and L-phenylalanyl-L-proline. The separation of the rotational

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service