Skip to Content
Merck
All Photos(1)

Documents

P3251

Sigma-Aldrich

Phe-Ala

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O3
CAS Number:
Molecular Weight:
236.27
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Phe-Ala,

Assay

≥98.0% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H](N)Cc1ccccc1)C(O)=O

InChI

1S/C12H16N2O3/c1-8(12(16)17)14-11(15)10(13)7-9-5-3-2-4-6-9/h2-6,8,10H,7,13H2,1H3,(H,14,15)(H,16,17)/t8-,10-/m0/s1

InChI key

MIDZLCFIAINOQN-WPRPVWTQSA-N

Gene Information

human ... SLC15A1(6564)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Döring et al.
The Journal of physiology, 497 ( Pt 3), 773-779 (1996-12-15)
l. Recently two genes have been identified by expression cloning that encode mammalian epithelial peptide transporters capable of translocating di- and tripeptides and selected peptidomimetics by stereoselective and rheogenic substrate-H+ cotransport. PepT1 from rabbit or human small intestine induces a
U Wenzel et al.
The American journal of physiology, 275(6 Pt 1), C1573-C1579 (1998-12-09)
The reabsorption of filtered di- and tripeptides as well as certain peptide mimetics from the tubular lumen into renal epithelial cells is mediated by an H+-coupled high-affinity transport process. Here we demonstrate for the first time H+-coupled uptake of dipeptides
J J Chen et al.
Peptides, 17(1), 31-37 (1996-01-01)
The purpose of present study was to determine whether peptidase activity affects the release of substance P (SP) and calcitonin gene-related peptide (CGRP) from spinal cord slices. When slices were exposed to various inhibitors of endopeptidase 24.11, the resting and
I Knütter et al.
Biochemistry, 40(14), 4454-4458 (2001-04-04)
This study was initiated to develop inhibitors of the intestinal H(+)/peptide symporter. We provide evidence that the dipeptide derivative Lys[Z(NO(2))]-Pro is an effective competitive inhibitor of mammalian PEPT1 with an apparent binding affinity of 5-10 microM. Characterization of the interaction
F Döring et al.
Biochemical and biophysical research communications, 232(3), 656-662 (1997-03-27)
The methylotrophic yeast Pichia pastoris was used for heterologous expression of the rabbit intestinal peptide transporter PepT1 and its functional characterization. PepT1 mediates the electrogenic transmembrane transport of di- and tripeptides and peptido-mimetics such as beta-lactam antibiotics and ACE-inhibitors. Functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service