Skip to Content
Merck
All Photos(1)

Key Documents

I8140

Sigma-Aldrich

IgG from rabbit serum

technical grade, ≥80% (SDS-PAGE), buffered aqueous solution

Synonym(s):

Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

conjugate

unconjugated

Quality Level

grade

technical grade

Assay

≥80% (SDS-PAGE)

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

IgG from rabbit serum has been used:
  • as a reference, standard or blocking agent in many immunoassays including dot blots, ELISA, Westerns, IEP, and immunodiffusion assays
  • in arthus reaction[1]
  • in chromatin immunoprecipitation[2]

Biochem/physiol Actions

Immunoglobulin G (IgG) regulates the immune responses such as phagocytosis and is also involved in the development of autoimmune diseases.[3] IgG1 regulates complement fixation in mice.[4]

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.2, containing 15 mM sodium azide

Preparation Note

Rabbit IgG is purified via fractionation from pooled normal rabbit serum

Analysis Note

Immunoelectrophoresis (IEP) is used to establish the identity of the Rabbit IgG. . A single major arc of precipitation in the gamma region is observed versus anti-rabbit whole serum and anti-rabbit IgG. Minor gamma arcs may be present versus anti-rabbit whole serum

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Matveeva et al.
Cell discovery, 2, 15046-15046 (2016-07-28)
Specialized chromatin structures such as nucleosomes with specific histone modifications decorate exons in eukaryotic genomes, suggesting a functional connection between chromatin organization and the regulation of pre-mRNA splicing. Through profiling the functional location of Poly (ADP) ribose polymerase, we observed
Frances Rena Bahjat et al.
Arthritis and rheumatism, 58(5), 1433-1444 (2008-04-29)
To assess whether R788, an orally bioavailable small molecule inhibitor of spleen tyrosine kinase (Syk)-dependent signaling, could modulate disease in lupus-prone (NZB x NZW)F1 (NZB/NZW) mice via inhibition of Fc receptor (FcR) and B cell receptor signaling. R788 was administered
Mhairi Laird et al.
Endocrinology, 158(4), 920-935 (2017-03-23)
Androgens are essential for the normal function of mature antral follicles but also have a role in the early stages of follicle development. Polycystic ovary syndrome (PCOS), the most common cause of anovulatory infertility, is characterized by androgen excess and
Aluya R Oseghale et al.
Blood cells, molecules & diseases, 79, 102345-102345 (2019-07-28)
Pharmacologic induction of fetal hemoglobin (HbF) is an effective strategy for treating sickle cell disease (SCD) by ameliorating disease severity. Hydroxyurea is the only FDA-approved agent that induces HbF, but significant non-responders and requirement for frequent monitoring of blood counts
Gregory P Priebe et al.
Journal of immunology (Baltimore, Md. : 1950), 181(7), 4965-4975 (2008-09-20)
In a murine model of acute fatal pneumonia, we previously showed that nasal immunization with a live-attenuated aroA deletant of Pseudomonas aeruginosa strain PAO1 elicited LPS serogroup-specific protection, indicating that opsonic Ab to the LPS O Ag was the most

Questions

  1. What is the concentration of this IgG?

    1 answer
    1. The protein concentration for I8410 is batch-dependent and ranges from 10.0 - 13.0 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service