Skip to Content
Merck
All Photos(1)

Documents

I5381

Sigma-Aldrich

IgG from mouse serum

reagent grade, ≥95% (SDS-PAGE), lyophilized powder

Synonym(s):

Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

unconjugated

grade

reagent grade

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

composition

Protein, ≥80%

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Mouse IgG is purified from normal mouse serum by fractionation and ion-exchange chromatography.

Application

Purified mouse IgG may be used as a reference antigen, standard, blocking agent, or coating protein in a variety of immunoassays including ELISA, dot immunobinding, Western immunoblotting, immunodiffusion, and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solid phase immunoadsorbents. Mouse IgG was used as standard in ELISA and chromatin immunoprecipitation assay. It was used to coat nano particles in the development of biosensors.

Physical form

Lyophilized from 0.01 M phosphate buffer, pH 7.2, with 0.015 M NaCl

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescent Biorecognition of Gold Nanoparticle-IgG Conjugates Self-Assembled on E-Beam Patterns
Powell T and Yoon JY
Biotechnol. Progress, 22, 106-110 (2006)
Lonnie J Lucas et al.
Biosensors & bioelectronics, 23(5), 675-681 (2007-09-18)
Double detection of microsphere light scattering and quantum dot emission was demonstrated for lab-on-a-chip immunoassay without using stationary support. We conjugated quantum dots (QDs) onto microspheres to enable multiplex assays as well as to enhance the limit of detection (LOD).
Paurvi Shinde et al.
Journal of leukocyte biology, 102(1), 57-69 (2017-04-23)
LPS is a powerful adjuvant, and although LPS-mediated TLR4 signaling has been exquisitely delineated, the in vivo mechanism of how TLR4 responses impact T cell priming is far less clear. Besides costimulation, TNF and type 1 IFN are dominant cytokines
C Rebouissou et al.
Blood, 91(12), 4727-4737 (1998-06-17)
Agonist antihuman gp130 transducer monoclonal antibodies (MoAbs) were used in SCID mice to grow myeloma cells whose survival and proliferation is dependent on gp130 transducer activation. The agonist anti-gp130 MoAbs neither bound to murine gp130 nor activated murine cells and
Roberto Salvatori et al.
European journal of endocrinology, 177(3), 257-266 (2017-06-22)
Mutations in the aryl hydrocarbon receptor-interacting protein ( Observational, inferential and experimental study, including: Nine European-origin, unrelated c.805_825dup-positive pedigrees (four familial, five sporadic from the UK, USA and France) included 16 affected (nine gigantism/four acromegaly/two non-functioning pituitary adenoma patients and

Related Content

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Access high-quality reagents for antibody workflows, including adjuvants, blocking reagents, conjugation kits, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service