Skip to Content
Merck
All Photos(1)

Key Documents

HT501850

Sigma-Aldrich

Formalin solution, neutral buffered, 10%

histological tissue fixative

Synonym(s):

Formaldehyde, Methanal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

biological source

synthetic

Quality Level

concentration

10% formalin (approx. 4% formaldehyde)

color

colorless

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formalin solution, neutral buffered, 10% may be used as a reagent for the primary fixation of tissues for diagnostic applications of an immunoperoxidase method in hematopathology.
Popular general purpose histological fixative.

Physical form

Neutral buffered solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaehwan Ahn et al.
Biomaterials, 268, 120599-120599 (2020-12-21)
Non-alcoholic fatty liver disease (NAFLD) has become a global pandemic. However, a pharmacological cure has not been approved for NAFLD treatment. The greatest barriers to the development of new treatments are the ambiguous criteria among the NAFLD stages and the
Diagnostic applications of an immunoperoxidase method in hematopathology.
Banks PM.
The Journal of Histochemistry and Cytochemistry, 27(8), 1192-1194 (1979)
Xiaozhen Liang et al.
PLoS pathogens, 7(9), e1002220-e1002220 (2011-09-21)
Human gammaherpesviruses are associated with the development of lymphoproliferative diseases and B cell lymphomas, particularly in immunosuppressed hosts. Understanding the molecular mechanisms by which human gammaherpesviruses cause disease is hampered by the lack of convenient small animal models to study
Caterina Nardella et al.
Science signaling, 2(55), ra2-ra2 (2009-01-30)
The mammalian target of rapamycin (mTOR) is a crucial effector in a complex signaling network commonly disrupted in cancer. mTOR exerts its multiple functions in the context of two different multiprotein complexes: mTOR complex 1 (mTORC1) and mTOR complex 2
Coralie Apfeldorfer et al.
Diagnostic pathology, 3 Suppl 1, S16-S16 (2008-08-05)
Historically, histopathology evaluation is performed by a pathologist generating a qualitative assessment on thin tissue sections on glass slides. In the past decade, there has been a growing interest for tools able to reduce human subjectivity and improve workload. Whole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service