Skip to Content
Merck
All Photos(1)

Key Documents

HT5011

Sigma-Aldrich

Formalin solution, neutral buffered, 10%

case of 48 × 15 mL, histological tissue fixative

Synonym(s):

Formaldehyde, Methanal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.47

packaging

case of 48 × 15 mL

concentration

10% formalin (approx. 4% formaldehyde)

color

colorless

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[O-]#[C+H2]

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde is both a non-coagulant and an additive fixative. Formaldehyde is known to penetrate tissue quickly, but the fixation process may take a long time to form the various cross links with tissue proteins. Formalin fixation is thought to form cross links between the aldehydes and the proteins, creating a gel, thus retaining cellular constituents in their in vivo relationship. Once properly fixed, the tissue should be able to withstand the subsequent stages of tissue processing or staining. Although there is no true universal fixative, 10% neutral buffered formalin (NBF) is perhaps the most commonly used fixative throughout the world for light microscopy and is a somewhat forgiving fixative. Of the dozens and dozens of available fixatives, it is hard to imagine a fixative more commonly used than 10% NBF. 10% neutral buffered formalin is well suited for large throughput laboratories, and requires a relatively short period of fixation, but can also be used for the long-term storage of tissue.

Application

10% neutral buffered formalin (NBF) is a popular general purpose histological fixative. It has been used to fix tissues, protein complexes and tumor samples.
Popular general purpose histological fixative.

Packaging

Case containers are polypropylene with white leak-proof lids and are partially prefilled with 10% NBF.

Physical form

Neutral buffered solution

Storage and Stability

Store tightly closed at room temperature. Do not freeze.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mangiferin induces islet regeneration in aged mice through regulating p16INK4a.
Wang H, et al.
International Journal of Molecular Medicine, 41(6), 3231-3242 (2018)
Biological characterization of a Stable Effector Functionless (SEFL) monoclonal antibody scaffold in vitro.
Liu L, et al.
The Journal of Biological Chemistry, 292(5), 1876-1883 (2017)
Nuclear GSK3β induces DNA double-strand break repair by phosphorylating 53BP1 in glioblastoma.
Yang Y, et al.
International Journal of Oncology (2018)
Parco M Siu et al.
BMC musculoskeletal disorders, 12, 58-58 (2011-03-10)
The molecular mechanism initiating deep pressure ulcer remains to be elucidated. The present study tested the hypothesis that the ubiquitin proteasome system is involved in the signalling mechanism in pressure-induced deep tissue injury. Adult Sprague Dawley rats were subjected to
Rune M Pedersen et al.
Frontiers in cellular and infection microbiology, 8, 16-16 (2018-02-17)
Most bacterial infections initiate at the mucosal epithelium lining the gastrointestinal, respiratory, and urogenital tracts. At these sites, bacterial pathogens must adhere and increase in numbers to effectively breach the outer barrier and invade the host. If the bacterium succeeds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service