Skip to Content
Merck
All Photos(1)

Documents

MINI67

Sigma-Aldrich

PKH67 Green Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling

Distributed for Phanos Technologies

Synonym(s):

Green PKH membrane mini labeling kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

packaging

pkg of 1 kit

manufacturer/tradename

Distributed for Phanos Technologies

storage condition

protect from light

technique(s)

flow cytometry: suitable

fluorescence

λex 490 nm; λem 502 nm (PKH67 dye)

application(s)

cell analysis
detection

detection method

fluorometric

shipped in

ambient

storage temp.

room temp

General description

PKH67 kit is for general cell membrane labeling. It has a longer aliphatic carbon tail than PKH1 and PKH2, two other green dyes previously described for in vitro and in vivo cell tracking. Based on the longer tail length, in-house studies have consistently shown reduced cell-cell transfer for PKH67 as compared to PKH2.
Slow loss of fluorescence has been observed in in vivo studies using PKH1 and PKH2. PKH67 may exhibit similar properties since this behavior appears to be characteristic of green cell linker dyes, but not red cell linker dyes. Correlation of in vitro cell membrane retention with in vivo fluorescence half life in non-dividing cells predicts an in vivo fluorescence half life of 10-12 days for PKH67. Other green cell linker dyes with similar half lives have been used to monitor in vivo lymphocyte and macrophage trafficking over periods of 1-2 months, suggesting that PKH67 will also be useful for in vivo tracking studies of moderate length.

Application

PKH67 green fluorescent cell linker mini kit for general cell membrane labeling has been used to stain:
  • exosomes
  • pericytes
  • UT-7/TPO cells

This kit is for general cell membrane labeling. Slow loss of fluorescence has been observed in in vivo studies using PKH1 and PKH2. PKH67 may exhibit similar properties since this behavior appears to be characteristic of green cell linker dyes, but not red cell linker dyes.

Linkage

For additional technical details on PKH and CellVue® Fluorescent Cell Linker Dyes including an extensive bibliography, please visit here.

Legal Information

Distributed for Phanos Technologies, Inc.
CellVue is a registered trademark of Phanos Technologies

Kit Components Only

Product No.
Description

  • Diluent C 10 mL

  • PKH67 Cell Linker in ethanol 0.1 mL

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exosomal MALAT1 derived from oxidized low-density lipoprotein-treated endothelial cells promotes M2 macrophage polarization
Huang C, et al.
Molecular Medicine Reports, 18(1), 509-515 (2018)
A microfluidic study of megakaryocytes membrane transport properties to water and dimethyl sulfoxide at suprazero and subzero temperatures
Tseng HY, et al.
Biopreservation and Biobanking, 9(4), 355-362 (2011)
Yuan Yang et al.
Experimental and therapeutic medicine, 20(6), 257-257 (2020-11-18)
Mesenchymal stem cell (MSC) infusion has become a novel therapeutic strategy for complex autoimmune diseases; however, few detailed studies have been performed to investigate the benefit and mechanism of MSC treatment on systemic sclerosis (SSc). The present study aimed to
Simone Moertl et al.
International journal of molecular sciences, 21(7) (2020-04-02)
Normal tissue toxicity is a dose-limiting factor in radiation therapy. Therefore, a detailed understanding of the normal tissue response to radiation is necessary to predict the risk of normal tissue toxicity and to development strategies for tissue protection. One component
Paul K Wallace et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 73(11), 1019-1034 (2008-09-13)
Cell-tracking reagents such as the green-fluorescent protein labeling dye CFSE and the red-fluorescent lipophilic membrane dye PKH26 are commonly used to monitor cell proliferation by flow cytometry in heterogeneous cell populations responding to immune stimuli. Both reagents stain cells with

Articles

PKH dyes are easy to use and achieve stable, uniform, and reproducible fluorescent labeling of live cells. PKH dyes are non-toxic membrane stains which produce high signal to noise ratio.

PKH dyes are easy to use and achieve stable, uniform, and reproducible fluorescent labeling of live cells. PKH dyes are non-toxic membrane stains which produce high signal to noise ratio.

PKH dyes are easy to use and achieve stable, uniform, and reproducible fluorescent labeling of live cells. PKH dyes are non-toxic membrane stains which produce high signal to noise ratio.

PKH dyes are easy to use and achieve stable, uniform, and reproducible fluorescent labeling of live cells. PKH dyes are non-toxic membrane stains which produce high signal to noise ratio.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service