Skip to Content
Merck
All Photos(1)

Key Documents

H4541

Sigma-Aldrich

HPI-4

≥98% (HPLC)

Synonym(s):

2,4-Dichloro-a-(3,4-dihydro-4-oxo-2(1H)-quinazolinylidene)-β-oxo-benzenepropanenitrile, Ciliobrevin A, Hedgehog Pathway Inhibitor 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H9Cl2N3O2
CAS Number:
Molecular Weight:
358.18
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white, off-white to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1ccc(c(Cl)c1)C(=O)\C(C#N)=C2\NC(=O)c3ccccc3N2

InChI

1S/C17H9Cl2N3O2/c18-9-5-6-10(13(19)7-9)15(23)12(8-20)16-21-14-4-2-1-3-11(14)17(24)22-16/h1-7,21H,(H,22,24)/b16-12+

InChI key

SESYPWCSIZUIAS-FOWTUZBSSA-N

Application

Hedgehog pathway inhibitor (HP1-4) has been used for the inhibition of dynein ATPase in Leishmania.

Biochem/physiol Actions

HPI-4 is a Hedgehog pathway inhibitor; ciliogenesis inhibitor. The Hedgehog (HH) signaling pathway is targeted for anti-cancer therapeutics. A key signaling molecule in HH pathway, Smoothened (Smo), has been the target of pharmacological intervention, which has resulted in multiple Smo antagonists including Cyclopamine. However, the oncogenic form of Smo is resistant to Cyclopamine, so the identification of inhibitors of downstream effectors, such as the transcription factors Gli1 and Gli2, is important. Four small molecules were identified to inhibit HH downstream of Smo, HPI-1, HPI-2, HPI-3, and HPI-4. HPI-4 blocked HH pathway activation by SAG, decreased Smo ciliary accumulation, decreased HH activity in a consitutively active HH cell line, and decreased Gli1 and Gli2, processing, and stability. HPI-4 inhibited proliferation of cerebellar granula neuron precursor cells, an important model of HH signaling. Cilia in HPI-4-treated cells were either truncated or absent. The mechanism of action of HPI-4 is hypothesized to be directly through perturbation of ciliogenesis, leading to disruption of Gli1/Gli2 activity, which is distinctly different from that of Cyclopamine.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Xiang et al.
Oncology reports, 32(4), 1622-1630 (2014-08-12)
Chondrosarcoma is a type of malignant bone tumor secreting cartilage-like matrix. In clinical treatment, there is no frequently used drug treatment option except for surgical resection. Hedgehog (HH) pathway is a classical signaling pathway that regulates normal cartilage cell development.
Ivonne Bernal et al.
Protein science : a publication of the Protein Society, 28(10), 1888-1901 (2019-08-09)
Translocation of virulence effector proteins through the type III secretion system (T3SS) is essential for the virulence of many medically relevant Gram-negative bacteria. The T3SS ATPases are conserved components that specifically recognize chaperone-effector complexes and energize effector secretion through the
Effect of inhibition of axonemal dynein ATPases on the regulation of flagellar and ciliary waveforms in Leishmania parasites
Mukhopadhyay AG and Dey CS
Molecular and Biochemical Parasitology, 225, 27 -237 (2018)
Ivonne Bernal et al.
Journal of molecular biology, 431(19), 3787-3803 (2019-07-10)
Many medically relevant Gram-negative bacteria use the type III secretion system (T3SS) to translocate effector proteins into the host for their invasion and intracellular survival. A multi-protein complex located at the cytosolic interface of the T3SS is proposed to act
Characterization of ciliobrevin A mediated dynein ATPase inhibition on flagellar motility of Leishmania donovani
Reddy GS, et al.
Molecular and Biochemical Parasitology, 214, 75 -781 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service