Skip to Content
Merck
All Photos(1)

Documents

GW21143

Sigma-Aldrich

Anti-AP-1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Activator protein 1, Anti-Transcription factor AP-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

manufacturer/tradename

Genway 15-288-21143

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JUN(3725)

Immunogen

Immunogen Sequence: GI # 4758616, sequence 1-240
Recombinant V-jun avian sarcoma virus 17 oncogene homolog

Application

Anti-AP-1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

c-Jun is a protein encoded by the JUN gene in humans. c-Jun in combination with c-Fos results in the formation of the AP-1 early response transcription factor. It is crucially involved in propagation of macrophage-driven inflammatory networks. AP1 may guide for the treatment decisions for endometriosis management and may provide a mechanistic endpoint for assessing efficacy of new agents aimed at curtailing inflammatory mechanisms that drive disease progression. It may possibly be utilized for the treatment of HPV-induced cervical cancers.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juling Lv et al.
International journal of clinical and experimental pathology, 11(8), 4221-4229 (2018-08-01)
Visible light has been previously demonstrated to induce retina damage and retinal ganglion cell (RGC) death through the mitochondrial pathway. The Fra-2 gene was detected as a transcriptional regulator, but whether it is involved in light-induced retina damage remains unclear.
Sutapa Mahata et al.
Nutrition and cancer, 65 Suppl 1, 88-97 (2013-05-25)
Plant products of Phyllanthus emblica Linn. are traditionally consumed for its immense nutritive and medicinal values. However, the molecular mechanism(s) by which it exerts it effects is less understood. In this study, we investigated mechanism of action of P. emblica
Mohamed A Salem et al.
Scientific reports, 12(1), 6578-6578 (2022-04-23)
Aging has become a concern for many people, especially women. Given that high-quality anti-aging products are of high cost; it has imperative to search for other economical sources. Essential oils are frequently used in cosmetics products due to a wide
Michael T Beste et al.
Science translational medicine, 6(222), 222ra16-222ra16 (2014-02-07)
Clinical management of endometriosis is limited by the complex relationship between symptom severity, heterogeneous surgical presentation, and variability in clinical outcomes. As a complement to visual classification schemes, molecular profiles of disease activity may improve risk stratification to better inform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service