Skip to Content
Merck
All Photos(1)

Key Documents

U7257

Sigma-Aldrich

Urotensin II human

≥97% (HPLC), synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H85N13O18S2
CAS Number:
Molecular Weight:
1388.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Quality Level

Assay

≥97% (HPLC)

form

lyophilized powder

technique(s)

calcium flux assay: suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@@H]1CSSC[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)[C@@H](N)CCC(O)=O)[C@@H](C)O)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc6ccc(O)cc6)C(=O)N1)C(O)=O

InChI

1S/C64H85N13O18S2/c1-33(2)52(64(94)95)75-61(91)48-32-97-96-31-47(73-59(89)46(29-51(82)83)72-62(92)49-17-11-25-77(49)63(93)53(34(3)78)76-54(84)40(66)22-23-50(80)81)60(90)70-43(26-35-12-5-4-6-13-35)56(86)71-45(28-37-30-67-41-15-8-7-14-39(37)41)58(88)68-42(16-9-10-24-65)55(85)69-44(57(87)74-48)27-36-18-20-38(79)21-19-36/h4-8,12-15,18-21,30,33-34,40,42-49,52-53,67,78-79H,9-11,16-17,22-29,31-32,65-66H2,1-3H3,(H,68,88)(H,69,85)(H,70,90)(H,71,86)(H,72,92)(H,73,89)(H,74,87)(H,75,91)(H,76,84)(H,80,81)(H,82,83)(H,94,95)/t34-,40+,42+,43+,44+,45+,46+,47+,48+,49+,52+,53+/m1/s1

InChI key

HFNHAPQMXICKCF-USJMABIRSA-N

Gene Information

human ... UTS2(10911)

Amino Acid Sequence

Glu-Thr-Pro-Asp-Cys-Phe-Trp-Lys-Tyr-Cys-Val [Disulfide Bridge: 5-10]

Application

Urotensin II human was used in agonist and antagonist preparation for calcium mobilization assay. The product can also be used as a test compound for studying the impact of dihydropyridine calcium antagonist amlodipine in human internal mammary artery and clinical implications.

Biochem/physiol Actions

A neuroendocrine peptide of the CRF family that was originally isolated from fish brain but is also found in mammalian brain and spinal motor neurons. It is a potent vasoconstrictor. It may be a mediator of stress-induced anorexia.
A neuroendocrine peptide of the CRF family; potent vasoconstrictor.
Human urotensin II is a vasoactive neuropeptide that belongs to CRF family and stimulates arterial smooth muscle contraction, actin stress fiber formation and proliferation via small GTPase RhoA and there downstream effector Rho-kinase activation.The product is also a selective and potent vasoconstrictor agent as well as an agonist for the orphan receptor GPR14.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Sauzeau et al.
Circulation research, 88(11), 1102-1104 (2001-06-09)
The aim of this work was to investigate the coupling of human urotensin II (hU-II) to RhoA activation and regulation of RhoA-dependent functions. The use of the Rho-kinase inhibitor Y-27632 and the development of a membrane-permeant RhoA inhibitor (TAT-C3) allowed
Xiao-Yan Bai et al.
The Annals of thoracic surgery, 92(2), 610-616 (2011-06-28)
Graft spasm in the internal mammary artery (IMA) may occur after coronary artery bypass grafting (CABG). We investigated the effect of human urotensin II (hU-II), a cyclic peptide hormone present in human blood and tissues, and the effect of vasodilators
Xiao-Yan Bai et al.
The Annals of thoracic surgery, 90(6), 1952-1957 (2010-11-26)
Graft spasm remains challenging in CABG (coronary artery bypass grafting) surgery. We investigated the inhibitory effect of a dihydropyridine calcium antagonist amlodipine on the vasoconstriction mediated by potassium chloride (KCl), human urotensin-II (hU-II), and U46619 in human internal mammary artery
Hong Xin et al.
Journal of biomolecular screening, 12(5), 705-714 (2007-06-01)
The no-wash calcium assay kits developed by Molecular Devices Corporation have greatly enhanced the throughput of cell-based calcium mobilization high-throughput screening (HTS) assays and enabled screening using nonadherent cells. The fluorescent imaging plate reader (FLIPR) Calcium 3 Assay Kit, optimal
R S Ames et al.
Nature, 401(6750), 282-286 (1999-09-28)
Urotensin-II (U-II) is a vasoactive 'somatostatin-like' cyclic peptide which was originally isolated from fish spinal cords, and which has recently been cloned from man. Here we describe the identification of an orphan human G-protein-coupled receptor homologous to rat GPR14 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service