Skip to Content
Merck
All Photos(3)

Key Documents

G9259

Sigma-Aldrich

α-Glucosidase from rice

greener alternative

Type V, ammonium sulfate suspension, 40-80 units/mg protein

Synonym(s):

α-D-Glucoside glucohydrolase, Maltase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type V

Quality Level

form

ammonium sulfate suspension

specific activity

40-80 units/mg protein

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch hydrolysis research. For more information see the article in biofiles.

Unit Definition

One unit will convert 1.0 μmole of maltose to 2.0 μmoles of D-glucose per min at pH 4.0 at 37 °C.

Physical form

Suspension in 2.8 M (NH4)2SO4 solution

Analysis Note

Protein determined by biuret.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Rugen et al.
Methods in molecular biology (Clifton, N.J.), 1795, 101-115 (2018-05-31)
The hydrolysis of starch is a key step in plant germination, which also has relevance in the malting and brewing processes for beer and spirit production. Gaps in knowledge about this metabolic process exist that cannot easily be addressed using
P R Flanagan et al.
The Biochemical journal, 173(2), 553-563 (1978-08-01)
Maltase-glucoamylase, a microvillous membrane ectoenzyme, was solubilized from rat intestinal mucosa by digestion with papain and subsequently purified to homogeneity with an overall yield of 10--20%. An antibody to the purified enzyme formed a single precipitin line in immunodiffusion experiments
Amphiphilic pig intestinal microvillus maltase/glucoamylase. Structure and specificity.
S H Sørensen et al.
European journal of biochemistry, 126(3), 559-568 (1982-09-01)
Rachel Mata et al.
Journal of natural products, 76(3), 468-483 (2013-02-13)
Type II-diabetes mellitus (TII-DM) has been regarded as one of the most important public health problems in all nations in the 21st century. Although allopathic therapies remain the most important for the initial management of TII-DM, herbal remedies have gained
Fushuang Li et al.
Organic letters, 15(3), 674-677 (2013-01-17)
Polyflavanostilbene A, a new flavanol-fused stilbene glycoside, was isolated from the rhizome of Polygonum cuspidatum. Its unusual structure, including its absolute stereochemistry, was determined by UV, IR, HRESIMS, and 1D and 2D NMR data and by the comparison of experimental

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service