Skip to Content
Merck
All Photos(1)

Key Documents

N1377

Sigma-Aldrich

4-Nitrophenyl α-D-glucopyranoside

chromogenic, ≥99%, powder or crystals

Synonym(s):

p-Nitrophenyl α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
Beilstein:
92212
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Nitrophenyl α-D-glucopyranoside, ≥99%

Assay

≥99%

form

powder or crystals

solubility

methanol: 20 mg/mL, clear to very slightly hazy, colorless to greenish-yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11-,12+/m1/s1

InChI key

IFBHRQDFSNCLOZ-ZIQFBCGOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl ǥ-D-glucopyranoside has been used as a substrate in an ǥ-glucosidase inhibition assay.

Biochem/physiol Actions

4-Nitrophenyl ǥ-D-glucopyranoside serves as a substrate for glycosidases. Upon cleavage, the substrate is converted to a yellow-colored product.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insight into anti-diabetic potential of Clematis apiifolia: glucose control in diabetes.
Zehra SA, et al.
Pakistan Journal of Agricultural Sciences null
Faryal Chaudhry et al.
Bioorganic chemistry, 82, 267-273 (2018-11-06)
Herein, substituted imidazole-pyrazole hybrids (2a-2n) were prepared via a multi component reaction employing pyrazole-4-carbaldehydes (1a-1d), ammonium acetate, benzil and arylamines as reactants. All the new compounds were characterized through their spectral and elemental analyses. Further these compounds were tested against
Jaegoo Kim et al.
World journal of microbiology & biotechnology, 34(11), 167-167 (2018-11-02)
Candida albicans is a major invasive pathogen, and the development of strains resistant to conventional antifungal agents has been reported in recent years. We evaluated the antifungal activity of 44 compounds against Candida strains. Magnoflorine showed the highest growth inhibitory
Sajjad Esmaeili et al.
Bioorganic chemistry, 88, 102972-102972 (2019-05-13)
Dipyridamole (DP) elevates cyclic Adenosine Monophosphate (cAMP) levels in platelets, erythrocytes, and endothelial cells and also blocks adenosine reuptake. It is used to dilate blood vessels in people with peripheral arterial disease and coronary artery disease (CAD). The flexible backbone
P S Unnikrishnan et al.
Pharmacognosy magazine, 11(Suppl 4), S511-S515 (2016-03-26)
In the continuing search for safe and efficient antidiabetic drug, marine algae become important source which provide several compounds of immense therapeutic potential. Alpha-amylase, alpha-glucosidase inhibitors, and antioxidant compounds are known to manage diabetes and have received much attention recently.

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service