Skip to Content
Merck
All Photos(1)

Key Documents

49290

Sigma-Aldrich

β-Glucosidase from almonds

lyophilized, powder, ≥4 U/mg

Synonym(s):

β-D-Glucoside glucohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

Quality Level

quality

lyophilized

specific activity

≥4 U/mg

mol wt

Mr ~135000

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

β-glucosidase is also used in the synthesis of glucosides and fucosides with various potential applications in pharmaceutical, cosmetic and detergent industries, hydrolytic removal of aglycone moiety from flavonoid and isoflavonoid glycosides, flavor enhancement of fruit juices and wine, and biosynthesis of oligosaccharides.

Biochem/physiol Actions

βglucosidase is involved in the hydrolysis of β-glycosidic bonds connecting carbohydrate residues in β-D-glycosides. They convert cellobiose and cellooligosaccharides produced by the endo and exoglucanases to glucose.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol glucose per minute at pH 5.0 and 37 °C (salicin as substrate)

Other Notes

Role of β-glucosidase in cellulose hydrolysis: Review; Enzymic hydrolysis of cellulose for fuel ethanol production by simultaneous saccharification and fermentation; Stereospecific attachment of carbohydrates to amino acid derivatives; Enzyme-catalyzed synthesis of alkyl β-D-glucosides in organic media

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G P Philippidis et al.
Biotechnology and bioengineering, 41(9), 846-853 (1993-04-15)
The biochemical conversion of cellulosic biomass to ethanol, a promising alternative fuel, can be carried out efficiently and economically using the simultaneous saccharification and fermentation (SSF) process. The SSF integrates the enzymatic hydrolysis of cellulose to glucose, catalyzed by the
G. Vic, and Thomas, D.
Tetrahedron Letters, 33, 4567-4567 (1992)
Yawei Zhan et al.
RSC advances, 12(26), 16942-16954 (2022-06-28)
In this study, the effect of the γ-valerolactone (GVL)/H2O pretreatment system on bamboo (Neosinocalamus affinis) for enzymatic hydrolysis and ethanol fermentation was investigated. The performance characterization of the pretreated bamboo substrates, including the chemical composition, the structural characteristics, and the
Turner, N.J., and Webberley, M.C.
Journal of the Chemical Society. Chemical Communications, 1349-1349 (1991)
Beta-Glucosidase: its role in cellulase synthesis and hydrolysis of cellulose.
J G Shewale
The International journal of biochemistry, 14(6), 435-443 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service