Skip to Content
Merck
All Photos(3)

Key Documents

G8270

Sigma-Aldrich

D-(+)-Glucose

≥99.5% (GC)

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

maize

Quality Level

Assay

≥99.5% (GC)

form

powder

optical activity

[α]20/D 52.7°, c = 10% (w/v) in water

technique(s)

gas chromatography (GC): suitable

color

white

mp

150-152 °C (lit.)

solubility

H2O: 2g + 15 mL (Tested)

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Glucose is a monosaccharide which is the main source of energy in the form of ATP for living organisms. The effect of adding glucose as a supplement to the infection media of Cryptosporidium parvum oocysts has been studied. The polymer-bound glucose template has been demonstrated to be useful in the multi-step parallel synthesis of various compounds.

Application

D-(+)-Glucose has been used as a standard for the estimation of total sugar in hydrolyzed starch by phenol-sulfuric acid method.
It has been used in the preparation of the following liquid media for culturing yeast cells:
  • CM glucose broth minus HIS (histidine)
  • CM glucose broth minus URA (uracil)
  • CM glucose broth minus TRP (tryptophan)

Preparation Note

The material may produce a yellowish solution if prepared at a higher concentration than 2 g + 15mL.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Principles of micro Sequential Injection Analysis in the Lab-on-Valve format and its Introduction into a Teaching Laboratory
Decuir MS, et al.
Journal of Flow Injection Analysis, 24(2), 103-103 (2007)
Batch and continuous biohydrogen production from starch hydrolysate by Clostridium species.
Chen SD, et al.
International Journal of Hydrogen Energy, 33(7), 1803-1812 (2008)
D-Glucose as a Pentavalent Chiral Scaffold.
Opatz T, et al.
European Journal of Organic Chemistry, 2003(8), 1527-1536 (2003)
Cindy Lalancette et al.
Applied and environmental microbiology, 76(2), 566-577 (2009-11-26)
The inactivation of Cryptosporidium oocysts is a main driver in the selection of water treatment disinfection strategies, and microbial risk analysis provides a sound basis for optimizing water treatment processes. U.S. Environmental Protection Agency method 1622/23 provides an estimate of
Adam T Waickman et al.
Nature communications, 10(1), 3666-3666 (2019-08-16)
Generating effective and durable T cell immunity is a critical prerequisite for vaccination against dengue virus (DENV) and other viral diseases. However, understanding the molecular mechanisms of vaccine-elicited T cell immunity remains a critical knowledge gap in vaccinology. In this

Protocols

Enzymatic Assay of Invertase

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

Measure hexokinase activity using a continuous spectrophotometric rate-determination assay at 340 nm, catalyzing D-hexose sugar phosphorylation using ATP.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service