Skip to Content
Merck
All Photos(1)

Key Documents

G8769

Sigma-Aldrich

D-(+)-Glucose solution

45% in H2O, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

D-Glucopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1281608
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

product line

BioXtra

form

solution

concentration

45% in H2O

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

General description

Glucose is the energy currency essential for physiological processes in the living cells. It serves as a precursor for glycoproteins, glycolipids and glycosaminoglycans.

Application

D-(+)-Glucose solution has been used as a medium supplement:
  • in Dulbecco′s Modified Eagle Medium (DMEM) for culturing Flp-In  293 cells
  • in dissection medium for coronal slices from rat brain
  • in neurobasal medium for culturing hippocampi cells

Provides the primary energy source for cell metabolism.

Biochem/physiol Actions

Glucose is a crucial component for the growth and maintenance of mammalian cells in in vitro conditions and is essential for productivity and quality. Its high metabolic turnover favors osteoclast differentiation. The levels of glucose modulate the migration and growth of tumor cells. HeLa cells display high mobility at low glucose levels.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High glucose and low specific cell growth but not mild hypothermia improve specific r-protein productivity in chemostat culture of CHO cells
Vergara M, et al.
Testing, 13(8), e0202098-e0202098 (2018)
Glucose: an energy currency and structural precursor in articular cartilage and bone with emerging roles as an extracellular signaling molecule and metabolic regulator
Mobasheri A
Frontiers in Endocrinology, 3, 153-153 (2012)
Investigation of the Influence of Glucose Concentration on Cancer Cells by Using a Microfluidic Gradient Generator without the Induction of Large Shear Stress
Ishida T, et al.
Micromachines, 7(9), 155-155 (2016)
An ex vivo method for evaluating the biocompatibility of neural electrodes in rat brain slice cultures
Koeneman BA, et al.
Journal of Neuroscience Methods, 137(2), 257-263 (2004)
Protein kinase C activation causes neurite retraction via cyclin D 1 and p60-katanin increase in rat hippocampal neurons
Korulu S, et al.
The European Journal of Neuroscience, 37(10), 1610-1619 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service