Skip to Content
Merck
All Photos(1)

Documents

G7134

Sigma-Aldrich

L-(−)-Galactose

≥99% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724622
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

SMILES string

OC[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4+,5-,6?/m0/s1

InChI key

WQZGKKKJIJFFOK-DHVFOXMCSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Galactose was shown to be a key intermediate in the molecular pathway of converting D-glucose to oxalic acid in Pistia stratiotes.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minoru Tomizawa et al.
Oncology letters, 14(1), 899-902 (2017-07-12)
Tissues surrounding hepatocellular carcinomas (HCCs) lack glucose. Hepatocyte selection medium (HSM) is deficient in glucose and is supplemented with galactose. HCC cells were cultured in HSM to investigate the stem cell markers α-fetoprotein (AFP) and cluster of differentiation 44 (CD44).
S E Keates et al.
Phytochemistry, 53(4), 433-440 (2000-03-24)
Axenic Pistia stratiotes L. plants were pulse-chase labeled with [14C]oxalic acid, L[1-14C]ascorbic acid, L-6-14C]ascorbic acid, D-[1-14C]erythorbic acid, L-[1-14C]galactose, or [1-14C]glycolate. Specific radioactivities of L-ascorbic acid (AsA), free oxalic acid (OxA) and calcium oxalate (CaOx) in labeled plants were compared. Samples
Jolene M Garber et al.
Communications biology, 3(1), 2-2 (2020-01-12)
Although the gastrointestinal pathogen Campylobacter jejuni was considered asaccharolytic, >50% of sequenced isolates possess an operon for L-fucose utilization. In C. jejuni NCTC11168, this pathway confers L-fucose chemotaxis and competitive colonization advantages in the piglet diarrhea model, but the catabolic
Patrick Hossler et al.
mAbs, 9(4), 715-734 (2017-04-05)
Protein glycosylation is arguably the paramount post-translational modification on recombinant glycoproteins, and highly cited in the literature for affecting the physiochemical properties and the efficacy of recombinant glycoprotein therapeutics. Glycosylation of human immunoglobulins follows a reasonably well-understood metabolic pathway, which
K Saeki et al.
Veterinary journal (London, England : 1997), 205(2), 288-296 (2015-05-16)
Metastases are associated with a poor prognosis for canine mammary gland tumours (CMGTs). Metastatic and non-metastatic clones were isolated previously from a single malignant CMGT cell line. The difference in metastatic potential between the two cell lines was hypothesised to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service