R4377
L-(+)-Ribose
≥98% (GC)
Synonym(s):
Aldehydo-L-ribose
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723084
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥98% (GC)
form
powder
color
white to colorless
mp
81-82 °C (lit.)
storage temp.
2-8°C
SMILES string
OC[C@@H]1OC(O)[C@@H](O)[C@H]1O
InChI
1S/C5H10O5/c6-1-2-3(7)4(8)5(9)10-2/h2-9H,1H2/t2-,3-,4-,5?/m0/s1
InChI key
HMFHBZSHGGEWLO-OWMBCFKOSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shahid Mahmood et al.
Enzyme and microbial technology, 135, 109510-109510 (2020-03-10)
An enzyme, l-ribose isomerase (l-RI), mostly catalyzes the isomerization of l-ribose and l-ribulose. These so-called rare sugars are essential for the treatment of cancer and other viral diseases. In the present study, l-ribose isomerase produced from a bacterium, Mycetocola miduiensis
Jeremie J Crawford et al.
Electrophoresis, 37(15-16), 2217-2225 (2016-06-09)
Single enzyme molecule assays on E. coli β-galactosidase were performed using a capillary electrophoresis-based method. Three types of assays were performed. The catalytic rate of 20 individual molecules was assayed in duplicate in the presence of 50 μM substrate. The
Xin-Xia Xue et al.
Bioresource technology, 268, 292-299 (2018-08-08)
In this study, organic acid could be used as co-catalyst for assisting solid acid SO42-/SnO2-argil to convert hemicellulose-derived D-xylose into furfural. The relationship between pKa of organic acid and turnover frequency (TOF) of co-catalysis with organic acid plus SO42-/SnO2-argil was
Wen-Chi Tseng et al.
Journal of biotechnology, 259, 168-174 (2017-07-29)
A putative l-ribose isomerase (EC 5.3.1.B3, l-RI) gene of Actinotalea fermentans ATCC 43279 was chemically synthesized, subcloned into pET-21b vector, and then overexpressed in Escherichia coli. After 0.5mM IPTG induction at 20°C for 20h, the recombinant l-RI was highly expressed
Sirinan Shompoosang et al.
Journal of bioscience and bioengineering, 121(1), 1-6 (2015-06-03)
We biosynthesized 6-deoxy-L-talose, 6-deoxy-L-sorbose, 6-deoxy-L-gulose, and 6-deoxy-L-idose, which rarely exist in nature, from L-fucose by coupling and sequential enzymatic reactions. The first product, 6-deoxy-L-talose, was directly produced from L-fucose by the coupling reactions of immobilized D-arabinose isomerase and immobilized L-rhamnose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service