G5291
L-α-Glycerophosphorylcholine
≥98%, powder
Synonym(s):
Choline Glycerophosphate, Choline alphoscerate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H20NO6P
Molecular Weight:
257.22
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
Recommended Products
Product Name
L-α-Glycerophosphorylcholine, from soybean, ≥98%, powder
biological source
soybean
Assay
≥98%
form
powder
solubility
water: 50 mg/mL, clear, colorless to very faintly yellow
storage temp.
−20°C
SMILES string
C[N](C)(C)CCOP(O)(=O)OC[C@@H](O)CO
InChI
1S/C8H21NO6P/c1-9(2,3)4-5-14-16(12,13)15-7-8(11)6-10/h8,10-11H,4-7H2,1-3H3,(H,12,13)
InChI key
XGPKIOAEPQZUED-UHFFFAOYSA-N
Related Categories
General description
L-α-Glycerophosphorylcholine is a phospholipid, which is a derivative of phosphatidylcholine.
Application
L-α-Glycerophosphorylcholine has been used to rescue choline auxotrophy. It has also been used for the synthesis of glycerophospholipids.
Biochem/physiol Actions
Increases inositol phosphate formation.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cholinergic precursors in the treatment of cognitive impairment of vascular origin: ineffective approaches or need for re-evaluation?
Parnetti L, et al.
Journal of the Neurological Sciences, 257(1-2), 264-269 (2007)
Cognitive improvement in mild to moderate Alzheimer's dementia after treatment with the acetylcholine precursor choline alfoscerate: a multicenter, double-blind, randomized, placebo-controlled trial
Moreno MDJM
Clinical Therapeutics, 25(1), 178-193 (2003)
Alzheimer's Disease: Current Research In Early Diagnosis (1990)
Overproduction of Phospholipids by the Kennedy Pathway Leads to Hypervirulence in Candida albicans
Tams RN, et al.
Frontiers in Microbiology, 10 (2019)
High-Throughput Plasma Lipidomics: Detailed Mapping of the Associations with Cardiometabolic Risk Factors
Huynh K, et al.
Cell Chemical Biology, 26(1), 71-84 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service