Skip to Content
Merck
All Photos(3)

Key Documents

G5050

Sigma-Aldrich

Sephadex® G-50

Superfine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

powder

technique(s)

affinity chromatography: suitable

swelling

1 g swells to 9-11 mL

bead size

20-50 μm

pH

2—12

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatrography and gel filtration chromatography.
Fractionation Range (MW)
Globular Proteins: 1,500 - 30,000
Dextrans: 500 - 10,000

Application

Sephadex®G-50 has been used to purify the polymerase chain reaction (PCR) sequenced products.

Other Notes

G5050-100G′s updated product number is GE17-0041-01

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vered Shkalim et al.
American journal of medical genetics. Part A, 149A(12), 2700-2705 (2009-11-19)
We present a family segregating for an autosomal dominant syndrome of hypotelorism, cleft palate/uvula, high-arched palate and mild mental retardation. Although these findings may suggest a form of holoprosencephaly, no holoprosencephaly was found on MRI of the proposita. Results of
Effect of probiotic treatment in broiler chicks on intestinal macrophage numbers and phagocytosis of Salmonella Enteritidis by abdominal exudate cells
Higgins SE, et al.
Poultry Science, 86(11), 2315-2321 (2007)
M L Jirout et al.
Human molecular genetics, 19(13), 2567-2580 (2010-04-10)
Understanding catecholamine metabolism is crucial for elucidating the pathogenesis of hereditary hypertension. Here we integrated transcriptional and biochemical profiling with physiologic quantitative trait locus (eQTL and pQTL) mapping in adrenal glands of the HXB/BXH recombinant inbred (RI) strains, derived from
S E Higgins et al.
Poultry science, 86(11), 2315-2321 (2007-10-24)
Previous data have indicated that a Lactobacillus-based probiotic culture (FM-B11) is efficacious in reducing Salmonella Enteritidis colonization within 24 h when administered within 1 h of challenge. We hypothesized that the innate immune system, specifically macrophages, may play a role
Nicole E Choquette et al.
Plant, cell & environment, 43(12), 3033-3047 (2020-08-28)
Ozone pollution is a damaging air pollutant that reduces maize yields equivalently to nutrient deficiency, heat, and aridity stress. Therefore, understanding the physiological and biochemical responses of maize to ozone pollution and identifying traits predictive of ozone tolerance is important.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service