Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sephadex
®
G-50
CAS Number:
9004-54-0
Products
Brand
Sigma-Aldrich (2)
Manufacturer
Physical Form
Product Line
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
G5050
Sephadex
®
G-50,
Superfine
Expand
View Pricing
GE17-0042-01
Sephadex
®
G-50,
Fine, Cytiva, pack of 100 g
Expand
View Pricing
S5897
Sephadex
®
G-50,
BioReagent, for molecular biology, DNA grade, fine
Expand
View Pricing
GE17-0041-01
Sephadex
®
G-50,
Superfine, Cytiva, pack of 100 g
Expand
View Pricing
GE17-0573-02
Sephadex
®
G-50,
DNA grade, Cytiva, pkg of (pack of 100 g)
Expand
View Pricing
S6022
Sephadex
®
G-50,
BioReagent, for molecular biology, DNA grade, medium
Expand
View Pricing
GE17-0042-02
Sephadex
®
G-50,
Fine, Cytiva, pack of 500 g
Expand
View Pricing
GE17-0043-01
Sephadex
®
G-50,
Medium, Cytiva, pack of 100 g
Expand
View Pricing
GE17-0573-01
Sephadex
®
G-50,
DNA grade, Cytiva, pack of 25 g
Expand
View Pricing
GE17-0043-02
Sephadex
®
G-50,
Medium, Cytiva, pack of 500 g
Expand
View Pricing
GE17-0045-02
Sephadex
®
G-50,
M DNA grade
Expand
View Pricing