Skip to Content
Merck
All Photos(2)

Key Documents

G5080

Sigma-Aldrich

Sephadex® G-50

Fine

Synonym(s):

Gel filtration resin, Sephadex G-50 Fine Medium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

technique(s)

affinity chromatography: suitable

matrix active group

phase

swelling

1 g swells to 9-11 mL

bead size

20-80 μm

application(s)

life science and biopharma

compatibility

Cytiva

General description

Sephadex G-50 Fine is a widely used gel filtration resin designed for desalting and buffer exchange of biomolecules with a molecular weight greater than 30,000. Its small bead size contributes to higher efficiency in these processes. Different types of Sephadex vary in terms of cross-linking, resulting in differences in swelling and molecular fractionation range. Sephadex G-50 is one of five G-types available, ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex G-50 comes in four different particle sizes, such as Coarse, Medium, Fine, and Superfine.
Sephadex® G-50 is a gel filtration medium used in affinity chromatography, protein chromatography, and gel filtration chromatography. Sephadex is a beaded gel prepared by crosslinking dextran with epichlorohydrin.
Fractionation Range (MW)
Globular Proteins: 1,500 - 30,000
Dextrans: 500 - 10,000

Application

Sephadex® G-50 has been used:
  • to remove the non-entrapped carboxyfluorescein (CF) from the liposome suspension
  • in the purification of monoclonal antibody (mAb) humanized MN-14 by centrifuged size-exclusion chromatography
  • in desalting the recombinant enzymes eluted in the fraction five and six

Sephadex® G-50 is suitable for use in:

  • the separation of low and high molecular weight molecules
  • affinity chromatography, protein chromatography, and gel filtration chromatography

Features and Benefits

  • Desalts, removes contaminants, and transfers to a new buffer in one step.
  • Suitable for DNA purification from small molecules using gel filtration.
  • Features a small bead size, resulting in shorter diffusion distances.
  • Considered a classic gel filtration resin.

  • Desalting with Sephadex is considered superior to dialysis because it saves time, has a low dilution factor, and recovers high activity even with minute amounts of sample

Other Notes

G5080-100G′s updated product number is GE17-0042-01
G5080-500G′s updated product number is GE17-0042-02

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical-scale radiolabeling of a humanized anticarcinoembryonic antigen monoclonal antibody, hMN-14, with residualizing 131I for use in radioimmunotherapy
Govindan SV, et al.
Journal of Nuclear Medicine, 46(1), 153-159 (2005)
Samuel Lopez-Nieves et al.
The New phytologist, 217(2), 896-908 (2017-10-11)
Diverse natural products are synthesized in plants by specialized metabolic enzymes, which are often lineage-specific and derived from gene duplication followed by functional divergence. However, little is known about the contribution of primary metabolism to the evolution of specialized metabolic
Factors affecting leakage of trapped solutes from phospholipid vesicles during thermotropic phase transitions
Hays LM, et al.
Cryobiology (2001)
L Zhang et al.
The EMBO journal, 14(2), 313-320 (1995-01-16)
Heme is a prosthetic group for numerous enzymes, cytochromes and globins, and it binds tightly, sometimes covalently, to these proteins. Interestingly, heme also potentiates binding of the yeast transcriptional activator HAP1 to DNA and inhibits mitochondrial import of the mammalian
L M Weiner et al.
Journal of immunology (Baltimore, Md. : 1950), 151(5), 2877-2886 (1993-09-01)
Bispecific monoclonal antibodies (BsmAb) with specificity for tumor Ag and effector cell trigger molecules have been shown to redirect the cytotoxicity of several peripheral blood mononuclear cell populations against relevant tumor. The BsmAb, 2B1, binds to the extracellular domain of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service