Skip to Content
Merck
All Photos(4)

Documents

G2128

Sigma-Aldrich

L-Glutamic acid hydrochloride

Synonym(s):

(S)-2-Aminoglutaric acid, (S)-2-Aminopentanedioic acid ammonium salt, Glu HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4 · HCl
CAS Number:
Molecular Weight:
183.59
Beilstein:
3565569
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

SMILES string

Cl.N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4.ClH/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);1H/t3-;/m0./s1

InChI key

RPAJSBKBKSSMLJ-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-glutamate is the major excitatory neurotransmitter in mammalian central nervous system (CNS). It interacts with membrane bound glutamate receptors.

Application

L-Glutamic acid hydrochloride has been used as a nutrient additive to study the effect of increased dissolved inorganic nitrogen (DIN) on the uptake of dissolved organic nitrogen (DON) and dissolved organic carbon (DOC), in a nitrogen-limited headwater forest stream.

Biochem/physiol Actions

L-glutamate plays key roles in development, learning, memory, plasticity and cognition. It plays crucial role in the pathogenesis of neuropathological diseases such as epilepsy, schizophrenia, stroke, ischemia, ALS (amyotrophic lateral sclerosis), Huntington′s disease and Parkinson′s disease. Glutamate is also responsible for the activation of long-term potentiation (LTP) and long-term depression (LTD). Induction of glutamate receptors plays a key role in the pathophysiology of migraine.
Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coupled cycling of dissolved organic nitrogen and carbon in a forest stream.
Brookshire E N J et al
Ecology, 86(9), 2487-2496 (2005)
B K Day et al.
Journal of neurochemistry, 96(6), 1626-1635 (2006-01-31)
L-glutamate (Glu) is the predominant excitatory neurotransmitter in the mammalian central nervous system. It plays major roles in normal neurophysiology and many brain disorders by binding to membrane-bound Glu receptors. To overcome the spatial and temporal limitations encountered in previous
Martina Curto et al.
The journal of headache and pain, 17, 47-47 (2016-05-01)
Activation of glutamate (Glu) receptors plays a key role in the pathophysiology of migraine. Both NMDA and metabotropic Glu receptors are activated or inhibited by metabolites of the kynurenine pathway, such as kynureninic acid (KYNA), quinolinic acid (QUINA), and xanthurenic
Wei Lu et al.
Neuron, 78(3), 433-439 (2013-05-15)
The cell-autonomous role of synaptic transmission in the regulation of neuronal structural and electrical properties is unclear. We have now employed a genetic approach to eliminate glutamatergic synaptic transmission onto individual CA1 pyramidal neurons in a mosaic fashion in vivo. Surprisingly
A new beginning for a broken mind: balancing neuregulin 1 reverses synaptic dysfunction.
Oscar Marín et al.
Neuron, 78(4), 577-579 (2013-05-31)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service