Skip to Content
Merck
All Photos(7)

Key Documents

G0751

Sigma-Aldrich

β-Glucuronidase from Helix pomatia

Type H-1, partially purified powder, ≥300,000 units/g solid

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type H-1

form

partially purified powder

specific activity

≥300,000 units/g solid

secondary activity

≥10,000 units/g solid sulfatase

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Glucuronidase Type H-1 from Helix pomatia is partially purified powder containing enzymes derived from the Roman snail. Many β-glucuronidases derived from mollusks also contain sulfatase activity.

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine

Biochem/physiol Actions

Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Quality

Many β-glucuronidases derived from mollusks also contain sulfatase activity.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 5.0 (30 min assay).
Sulfatase Unit Definition: One unit of sulfatase will hydrolyze 1.0 μmole p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Briskey et al.
European journal of nutrition, 58(5), 2087-2097 (2018-07-06)
Curcumin has been shown to deliver protective effects against numerous degenerative conditions associated with high levels of inflammation and oxidative stress. Owing to its poor bioavailability when delivered orally, it is difficult to deliver a high concentration therapeutic dose. LipiSperse®
Myriam Richelle et al.
The Journal of nutrition, 132(9), 2587-2592 (2002-09-11)
We investigated whether the bioavailability of isoflavones could be enhanced by enzymatic hydrolysis of glycosides to aglycones before consumption of a nonfermented soy food. Two drinks were formulated with an enriched isoflavone extract from soy germ (Fujiflavone P10), one of
M Richelle et al.
European journal of clinical nutrition, 53(1), 22-26 (1999-02-27)
To evaluate the plasma kinetics in man of epicatechin from black chocolate. An intervention study with 8 volunteers. Each served as his own control. Theobromine was used as control marker of the chocolate intake. Metabolic Unit, Nestle Research Center, Vers-chez-les-Blanc
Kilia Y Liu et al.
The Journal of nutrition, 150(2), 222-230 (2019-09-10)
Vitamin E α-, γ-, or δ-tocopherol (αT, γT, δT) and γ- or δ-tocotrienol (γTE, δTE) are metabolized to hydroxychromanols and carboxychromanols including 13'-carboxychromanol (13'-COOH), 11'-COOH, and carboxyethyl hydroxychroman (CEHC), some of which have unique bioactivities compared with the vitamers. However
Shu-Shu Zhong et al.
ACS omega, 6(12), 8163-8170 (2021-04-06)
Arylsulfatase and β-glucuronidase are two important enzymes in humans, which play an important role in the dynamic equilibrium of steroidal estrogens. This work probably for the first time reported that hydrogen peroxide (H2O2), hypochlorite, and peracetic acid (PAA) could effectively

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Protocols

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service