Skip to Content
Merck
All Photos(8)

Key Documents

G0876

Sigma-Aldrich

β-Glucuronidase from Helix pomatia

Type H-2, aqueous solution, ≥85,000 units/mL

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type H-2

form

aqueous solution

specific activity

≥85,000 units/mL

secondary activity

≤7,500 units/mL sulfatase

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine

Biochem/physiol Actions

Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Quality

Many β-glucuronidases derived from mollusks also contain sulfatase activity.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 5.0 (30 min assay).
Sulfatase Unit Definition: One unit of sulfatase will hydrolyze 1.0 μmole p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C.

Physical form

Aqueous solution in ~1.0 M ammonium sulfate with 3 mM sodium azide as preservative.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takemi Akahane et al.
International journal of molecular sciences, 22(21) (2021-11-14)
Equol is a metabolite of daidzein, a major soybean isoflavone with estrogenic and antioxidant activities. As the production of equol depends on the presence of certain members of the intestinal microflora, not all individuals can produce equol. We examined the
Ke Wang et al.
Microbiology (Reading, England), 150(Pt 10), 3327-3339 (2004-10-08)
In Saccharomyces cerevisiae, PRS genes comprise a family of five paralogous genes. Previously, it has been shown that in the cell the gene products are organized into two interacting complexes, one of which is a heterodimer and the other a
Aynun Nahar Begum et al.
The Journal of nutrition, 134(1), 120-127 (2004-01-06)
The mammalian lignans enterolactone (ENL) and enterodiol, commonly found in human plasma and urine, are phytoestrogens that may contribute to the prevention of breast cancer and coronary heart disease. They are formed by the conversion of dietary precursors such as
Han-Hsing Tsou et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 28(5), 954-962 (2019-03-08)
Cigarette smoking (CS) and betel quid (BQ) chewing are two known risk factors and have synergistic potential for the development of oral squamous cell carcinoma (OSCC) in Taiwan. The p53 mutation characteristics in OSCC (G to A or G to
Tien-Jen Lin et al.
International journal of environmental research and public health, 15(7) (2018-07-11)
The measurement of polycyclic aromatic hydrocarbons (PAH) in ambient air is quite difficult to perform. Using urine biomarkers of PAH such as 2-naphthol is one approach to this problem. This study explored the association between urine 2-naphthol levels and allergic

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Protocols

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Enzymatic assay protocol for β-Glucuronidase from Helix Pomatia and Bovine Liver aids laboratory personnel in following procedures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service