Skip to Content
Merck
All Photos(1)

Key Documents

F5766

Sigma-Aldrich

Monoclonal Anti-Folic Acid antibody produced in mouse

clone VP-52, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

VP-52, monoclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:500

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal Anti-Folic Acid (mouse IgG2b isotype) is derived from the hybridoma produced from the fusion of mouse myeloma cells and splenocytes of an immunized mouse. The normal range for folate in serum is 3-13 ng/ml.

Specificity

Monoclonal Anti-Folic Acid antibody binds specifically to folate and detects an epitope present on both folic acid and its biologically active analog 5-methyltetrahydrofolic acid (5MTHFA). The product reacts with free folate, folate conjugated to a carrier such as KLH or BSA, and folic acid in human plasma or serum bound to the endogenous Folate Binder. The antibody does not cross-react with tetrahydrofolic acid, folinic acid or dihydrofolic acid.

Immunogen

5-Methyltetrahydrofolic acid (5MTHFA) conjugated to KLH

Application

Monoclonal Anti-Folic Acid antibody is suitable for use in western blot (diluted 1:1000).

Biochem/physiol Actions

Folic acid (folate) is a vitamin that regulates one-carbon transfer during the conversion of homocysteine to methionine. Methionine is a precursor of S-adenosylmethionine, which is the main methyl donor for methylation reactions. Thus, folic acid is essential for regulation of DNA methylations and for epigenetic modifications that modulate gene expression. Folate deficiency has been associated with a wide range of disorders including colorectal cancers, depression, neural tube defects and cerebral dysfunctions
Folic acid is essential for normal growth of mammalian cells. Folic acid deficiencies are common in pregnant women, alcoholics, those whose diets do not include raw fruits and vegetables, and people with structural or functional damage to the small intestine. Vitamin B12 and folic acid are metabolically interrelated.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linglei Jiang et al.
Biochimica et biophysica acta. General subjects, 1865(4), 129763-129763 (2020-10-17)
Extracellular vesicles (EVs) are nanoparticles which are released by cells from all three domains of life: Archaea, Bacteria and Eukarya. They can mediate cell-cell communication by transferring cargoes such as proteins and nucleic acids between cells. EVs receive great interest
Clinical utility of serum folate measurement in tertiary care patients: Argument for revising reference range for serum folate from 3.0 ng/mL to 13.0 ng/mL
Singh G, et al.
practical laboratory medicine, 1, 35-35 (2015)
Giuseppe Destito et al.
Chemistry & biology, 14(10), 1152-1162 (2007-10-27)
Cowpea mosaic virus (CPMV) is a well-characterized nanoparticle that has been used for a variety of nanobiotechnology applications. CPMV interacts with several mammalian cell lines and tissues in vivo. To overcome natural CPMV targeting and redirect CPMV particles to cells
Young-In Kim
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 13(4), 511-519 (2004-04-07)
Epidemiological, clinical, and animal studies collectively indicate that dietary folate intake and blood folate levels are inversely associated with colorectal cancer risk. Folate plays an essential role in one-carbon transfer involving remethylation of homocysteine to methionine, which is a precursor
Periconceptional folate deficiency and implications in neural tube defects.
Safi, J., et al.
Journal of Pregnancy, doi: 10-doi: 10 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service