Skip to Content
Merck
All Photos(2)

Documents

F2552

Sigma-Aldrich

Formestane

solid

Synonym(s):

4-Hydroxyandrost-4-ene-3,17-dione, CGP-32349

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

chloroform: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

application(s)

forensics and toxicology
veterinary

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=C(O)C(=O)CC[C@]34C)[C@@H]1CCC2=O

InChI

1S/C19H26O3/c1-18-10-8-15(20)17(22)14(18)4-3-11-12-5-6-16(21)19(12,2)9-7-13(11)18/h11-13,22H,3-10H2,1-2H3/t11-,12-,13-,18+,19-/m0/s1

InChI key

OSVMTWJCGUFAOD-KZQROQTASA-N

Gene Information

human ... CYP19A1(1588)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Aromatase inhibitor used as an anti-cancer agent against estrogen-dependent tumors.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Mauri et al.
Journal of the National Cancer Institute, 98(18), 1285-1291 (2006-09-21)
Aromatase inhibitors and inactivators have been extensively tested in patients with advanced breast cancer, but it is unclear whether they offer any survival benefits compared with standard hormonal treatment with tamoxifen or progestagens. We performed a meta-analysis of randomized controlled
P E Lønning et al.
The Journal of steroid biochemistry and molecular biology, 77(1), 39-47 (2001-05-19)
Formestane (Lentaron(R), 4-hydroxyandrostenedione) is a steroidal aromatase inhibitor used for treatment of advanced breast cancer. Clinically, it is administered as a depot form once fortnightly by intramuscular (i.m.) injection. To investigate the pharmacokinetics, bioavailability and metabolism of the drug, seven
P E Lønning
Breast cancer research and treatment, 49 Suppl 1, S45-S52 (1998-10-31)
Steroidal aromatase inhibitors like formestane and exemestane are useful drugs for endocrine treatment of postmenopausal breast cancer. In addition, these drugs should be considered valuable probes to explore the biology of breast cancer with particular emphasis on possible relations between
Xin Wang et al.
Cancer research, 66(21), 10281-10286 (2006-11-03)
Using Western blot as the major technique, we studied the effects of the three Food and Drug Administration (FDA)-approved aromatase inhibitors (AI) on aromatase protein stability in the aromatase-overexpressing breast cancer cell line MCF-7aro. We have found that exemestane treatment
Adam T Cawley et al.
Rapid communications in mass spectrometry : RCM, 22(24), 4147-4157 (2008-11-26)
Studies have shown that the administration of androstenedione (ADIONE) significantly increases the urinary ratio of testosterone glucuronide to epitestosterone glucuronide (T/E) - measured by gas chromatography/mass spectrometry (GC/MS) - in subjects with a normal ( approximately 1) or naturally high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service