Skip to Content
Merck
All Photos(4)

Key Documents

P0409

Sigma-Aldrich

Prostaglandin E2

synthetic, powder, BioReagent, suitable for cell culture

Synonym(s):

(5Z,11α,13E,15S)-11,15-Dihydroxy-9-oxoprosta-5,13-dienoic acid, Dinoprostone, PGE2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
Beilstein:
4709356
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
42020658
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic
form:
powder
Assay:
≥93% (HPLC)
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

product line

BioReagent

Assay

≥93% (HPLC)

form

powder

potency

0.25-100 ng/mL

technique(s)

cell culture | mammalian: suitable

solubility

acetone: 10 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@@H]1CC([C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/[C@@H](O)CCCCC)=O

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,19,21,23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,19+/m0/s1

InChI key

XEYBRNLFEZDVAW-ARSRFYASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prostaglandin E2 is a lipid that is obtained from arachidonic acid. Prostaglandin hormone is synthesized by the activity of cycloxygenase-2. Prostaglandin E2 controls inflammatory responses and vascular smooth muscle activity.[1]

Application

Prostaglandin E2 has been used as a α-MEM medium supplement for the co-culture of bone marrow macrophages and differentiated osteoblast.[2] It has also been used in the characterization of cells and in vitro decidualization.[3]

Biochem/physiol Actions

Most biologically active prostaglandin.
Most biologically active prostaglandin. PGE2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation; regulates adenylyl cyclase. Tumor cells that over-express cyclooxygenase 2 display increased invasiveness, angiogenesis, and resistance to apoptosis that may be due to the PGE2-induced expression of angiogenic factors and stabilization of the anti-apoptotic protein, survivin.The effect of PGE2 on the immune system is mixed. It inhibits T cell activation in vitro, suggesting it is an immunosuppressant. However, in vivo, it appears to effect expansion of the Th17 subset and differentiation of the Th1 subset of T helper cells, marking it as an immunoactivator.
Prostaglandin E2 is a signaling molecule produced by activated platelets. The release of PGE2 by activated platelets is part of a mechanism by which activated platelets utilize adjacent erythrocytes to help in clot formation. This product was shown to lower the filterability of human erythrocytes by approximately 30% at a concentration of 10-10sup M and also caused a reduction in mean cell volume by about 10%. The cause of cell shrinkage was the induction of a PGE2- stimulated K+ efflux pathway leading to rapid loss of cellular K+ ions. This loss was shown to be Ca2+dependent. PGE2 has been shown to stimulate the production of interleukin-6 (IL-6) by neonatal mouse parietal bones. After 6 hours in culture, cells stimulated with 10-8sup M PGE2 produced significantly more IL-6 than controls. The pyrogenic activity of PGE2 was not inhibited by dexamethasone, unlike prostaglandin F2α.

Most biologically active prostaglandin. PGE2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation; regulates adenylyl cyclase. Tumor cells that over-express cyclooxygenase 2 display increased invasiveness, angiogenesis, and resistance to apoptosis that may be due to the PGE2-induced expression of angiogenic factors and stabilization of the anti-apoptotic protein, survivin.

Physical form

powder -20 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analyses of bone modeling and remodeling using in vitro reconstitution system with two-photon microscopy
Hikita A, et al.
Bone, 76, 5-17 (2015)
Decidualization of the canine uterus: From early until late gestational in vivo morphological observations, and functional characterization of immortalized canine uterine stromal cell lines
Graubner F R, et al.
Reproduction in domestic animals = Zuchthygiene, 52, 137-147 (2017)
Potential use of G protein-coupled receptor-blocking monoclonal antibodies as therapeutic agents for cancers
International Review of Cell and Molecular Biology, 297, 45-81 (2012)
José L Maravillas-Montero et al.
Journal of immunology (Baltimore, Md. : 1950), 194(1), 29-33 (2014-11-21)
Chemokines are chemotactic cytokines that direct the traffic of leukocytes and other cells in the body. Chemokines bind to G protein-coupled receptors expressed on target cells to initiate signaling cascades and induce chemotaxis. Although the cognate receptors of most chemokines
Takayuki Nakagawa
Hearing research, 276(1-2), 27-33 (2011-02-08)
Prostaglandins are one of the major groups of chemical mediators in the mammalian body. Among prostaglandins, prostaglandin E2 (PGE2) is the most abundant prostanoid in humans and involved in regulating many different fundamental biological functions. PGE2 signaling is mediated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service