Skip to Content
Merck
All Photos(5)

Key Documents

H7904

Sigma-Aldrich

(Z)-4-Hydroxytamoxifen

≥98% (HPLC), powder, estrogen receptor modulator

Synonym(s):

4OH-Tamoxifen, (Z)-4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, (Z)-4-OHT, trans-4-Hydroxytamoxifen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29NO2
CAS Number:
Molecular Weight:
387.51
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Product Name

(Z)-4-Hydroxytamoxifen, ≥98% Z isomer

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

solubility

methanol: 10 mg/mL
ethanol: 20 mg/mL (with heating)

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

CC\C(c1ccccc1)=C(/c2ccc(O)cc2)c3ccc(OCCN(C)C)cc3

InChI

1S/C26H29NO2/c1-4-25(20-8-6-5-7-9-20)26(21-10-14-23(28)15-11-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h5-17,28H,4,18-19H2,1-3H3/b26-25-

InChI key

TXUZVZSFRXZGTL-QPLCGJKRSA-N

General description

4-Hydroxytamoxifen (4-OHT) is a metabolite of the antiestrogen tamoxifen in humans and other mammals. Both the Z (trans) and E (cis) 4-OHT isomers exhibit antiestrogenic properties in immature rats. Studies on the structure-function relationships of fixed ring systems have revealed that the trans isomer is a potent antiestrogen, while the cis isomer is relatively weaker (about 100 times less potent) as an antiestrogen in T47D breast cancer cells in vitro. 4-OHT binds to estrogen receptors (ER) and estrogen-related receptors (ERR), exerting both estrogenic and anti-estrogenic effects. This compound is a cell-permeable, selective estrogen receptor modulator (SERM). Compared to tamoxifen and its other metabolites, 4-OHT demonstrates a higher affinity for binding to estrogen receptors, resulting in 50 to 100-fold greater potency in inhibiting cell proliferation in normal human breast cells and breast cancer cell lines in culture. Moreover, 4-OHT has been found effective in inhibiting the growth of these cells in the absence of estrogen when cell proliferation was induced by insulin or epidermal growth factor.

Application

(Z)-4-Hydroxytamoxifen has been used:
  • as a media supplement to study the cell viability by WST-1 Assay
  • for inducing deletion of transforming growth factor-β (TGF-β gene in mice
  • to induce Cre recombinase activity in vitro.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microglia-Specific Expression of Olfml3 Is Directly Regulated by Transforming Growth Factor ?1-Induced Smad2 Signaling
Neidert N, et al.
Frontiers in Immunology, 9, 1728-1728 (2018)
Keratinocyte-derived TGF? is not required to maintain skin immune homeostasis
Yang Y, et al.
Journal of Dermatological Science, 94(2), 290-297 (2019)
Tamoxifen and hydroxytamoxifen as intramembraneous inhibitors of lipid peroxidation. Evidence for peroxyl radical scavenging activity
Cust'odio JBA, et al.
Biochemical Pharmacology, 47, 1989-1998 (1994)
Formulation of anti-miR-21 and 4-hydroxytamoxifen co-loaded biodegradable polymer nanoparticles and their antiproliferative effect on breast cancer cells
Devulapally R, et al.
J. Mol. Pharmacol., 12, 2080-2092 (2015)
Pei-Shan Hou et al.
Nature communications, 10(1), 3581-3581 (2019-08-10)
The bimodal requisite for a genetic program and external stimuli is a key feature of sensory circuit formation. However, the contribution of cell-intrinsic codes to directing sensory-specific circuits remains unknown. Here, we identify the earliest molecular program that preselects projection

Articles

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Modulation of homology-directed repair (HDR) within the context of CRISPR-genome editing has led to the identification of small molecules that enhance CRISPR-mediated HDR efficiency in various cell types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service