Skip to Content
Merck
All Photos(1)

Key Documents

E-075

Supelco

Estrone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC12CCC3C(CCc4cc(O)ccc34)C1CCC2=O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1

InChI key

DNXHEGUUPJUMQT-CBZIJGRNSA-N

Gene Information

human ... ESR1(2099)

General description

A precursor of estradiol, estrone is monitored in serum or plasma by LC-MS/MS or GC/MS to assess and diagnose developmental abnormalities, enzyme inactivity or deficiency, polycystic ovary syndrome (PCOS), or hormone replacement therapy in women.

Application


  • Fluconazole solution for yeast infection treatment: Fluconazole is extensively utilized for its efficacy against yeast infections, specifically vulvovaginal candidiasis. It operates by inhibiting the synthesis of ergosterol, crucial for fungal cell membrane integrity, thereby exerting its antifungal effects (Picheta et al., 2024).

  • Antifungal medication in systemic candidiasis: Fluconazole solution is critical in treating systemic candidiasis, offering a potent antifungal mechanism that disrupts the fungal cell membrane′s ergosterol content, essential for maintaining fungal cell viability (Dettlaff et al., 2024).

  • Fluconazole in vitro antifungal activity assay development: Fluconazole is utilized in the development of in vitro antifungal activity assays, which are crucial for screening potential antifungal agents and assessing their efficacy against various fungal pathogens (Teba et al., 2024).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emma M Ferguson et al.
Water research, 47(4), 1604-1615 (2013-01-12)
Estuaries are often the final repositories for aquatic pollutants but how estuarine hydrology influences the availability of marine- and freshwater-derived pollutants is not well understood, particularly for micro-pollutants such as endocrine disrupting chemicals. To address this knowledge gap, this study
Susannah J Stanway et al.
The oncologist, 12(4), 370-374 (2007-05-02)
Inhibitors of steroid sulfatase are being developed as a novel therapy for hormone-dependent breast cancer in postmenopausal women. Data suggest that steroid sulfatase (STS) activity is much higher than aromatase activity in breast tumors and high levels of STS mRNA
Kiyoshi Ito
The Tohoku journal of experimental medicine, 212(1), 1-12 (2007-04-28)
Hormone replacement therapy (HRT) has become available over the past few decades, but the risk of breast cancer with HRT remains controversial. The Women's Health Initiative Study has recently demonstrated that women receiving estrogen plus progestin (HRT) have an increased
Ghada N Farhat et al.
Journal of the National Cancer Institute, 105(19), 1496-1503 (2013-09-18)
Although high endogenous sex hormone levels and estrogen plus progestin (E+P) therapy are associated with increased breast cancer risk, it is unknown whether pretreatment levels of sex hormones modify E+P effect on breast cancer. We conducted a nested case-control study
Eva Fetter et al.
Aquatic toxicology (Amsterdam, Netherlands), 154, 221-229 (2014-06-14)
Xenoestrogens may persist in the environment by binding to sediments or suspended particulate matter serving as long-term reservoir and source of exposure, particularly for organisms living in or in contact with sediments. In this study, we present for the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service